The recommended shelf life for this product is 1 year from date of receipt.
Application Note: For ICC, formaldehyde fixation is recommended. Permeabilization with Triton-X 100 is recommended for formaldehyde-fixed cells.
This gene encodes a protein that is clearly involved in kinetochore function although an exact role is not known. It interacts with ZW10, another kinetochore protein, possibly regulating the association between ZW10 and kinetochores. The encoded protein localizes to prophase kinetochores before ZW10 does and it remains detectable on the kinetochore until late anaphase. It has a uniform distribution in the cytoplasm of interphase cells. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: human ZW10 interacting protein-1; SNAP25 interacting protein of 30 kDa; ZW10 interactor; ZW10 interactor, kinetochore protein; ZW10-interacting protein 1; Zwint-1
Gene Aliases: HZwint-1; KNTC2AP; SIP30; ZWINT; ZWINT1
UniProt ID: (Human) O95229
Entrez Gene ID: (Human) 11130
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support