Component of the ATAC complex, a complex with histone acetyltransferase activity on histones H3 and H4.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATAC component 1 homolog; DKFZP564I052; zinc finger, ZZ domain containing 3; zinc finger, ZZ-type containing 3; ZZ-type zinc finger-containing protein 3
Gene Aliases: 3110065C23Rik; 6430567E01Rik; AA408566; AI481057; ATAC1; AW556059; R75514; ZZZ3
UniProt ID: (Human) Q8IYH5, (Mouse) Q6KAQ7
Entrez Gene ID: (Human) 26009, (Mouse) 108946
Molecular Function:
transcription factor
zinc finger transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support