Applications | Tested Dilution | Publications |
---|---|---|
ELISA (ELISA) |
5-10 µg/mL | - |
Product Specifications | |
---|---|
Species Reactivity |
Chemical |
Host/Isotype |
Sheep / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Zilpaterol(N-imidazo)-BSA |
Conjugate |
Unconjugated |
Form |
Liquid |
Purification |
purified |
Storage buffer |
PBS |
Contains |
0.09% sodium azide |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
RRID |
AB_1091155 |
PA1-75517 specifically detects Zilpaterol.
PA1-75517 has been successfully used in ELISA applications.
The PA1-75517 immunogen is Zilpaterol(N-imidazo)-BSA.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: C14H19N3O2
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support