Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Immunogen sequence: DKNVIFSPL SISTALAFLS LGAHNTTLTE ILKGLKFNLT ETSEAEIHQS FQHLLRTLNQ SSDELQLSMG NAMFVKEQLS LLDRFTEDAK RLYGSEAFAT DFQDSAAAKK LINDYVKNGT RGKITDLIKD LDSQTMMVLV NYIFFKAKWE MPFDPQDTHQ SRFYLSKKKW VMVPMMSLHH LTIPYFRDEE LSCTVVELKY TGNASALFIL PDQDKMEEVE AMLLPETLKR WRDSLEFREI GELYLPKFSI SRDYNLNDIL LQLGIEEAFT SKADLSGITG ARNLAVSQVV HKAVLDVFEE GTEASAATAV KITLLSALVE TRTIVRFNRP FLMIIVPTDT QNIFFMSKVT NPKQA (70-423 aa encoded by B C010530)
SERPINA encodes Alpha 1-ACT. It is an early-stage acute-phase plasma protein and a serpin that preferentially inactivates chymotrypsin, cathepsin G, and chymase. Alpha-1-ACT, a serine protease inhibitor, is tightly associated with amyloid plaques in Alzheimer's disease (AD) and in normal aged human and monkey brain. Regulation of the serine proteases and serine protease inhibitors plays an important role in neuromuscular differentiation. Prostate specific antigen (PSA), a chymotrypsin-like serine protease, is predominantly complexed to Alpha-1-ACT.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ACT; Alpha-1-antichymotrypsin; Alpha-1-antichymotrypsin His-Pro-less; Cell growth-inhibiting gene 24/25 protein; Growth inhibiting protein 24; Growth inhibiting protein 25; growth-inhibiting protein 24; growth-inhibiting protein 25; MGC88254; Serine (or cysteine) proteinase inhibitor clade A member 3; serine (or cysteine) proteinase inhibitor, clade A, member 3; Serpin A3; Serpin peptidase inhibitor clade A (alpha 1 antiproteinase antitrypsin) member 3; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 3
Gene Aliases: AACT; ACT; GIG24; GIG25; SERPINA3
UniProt ID: (Human) P01011
Entrez Gene ID: (Human) 12
Molecular Function: protease inhibitor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support