Invitrogen
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:200 | - |
Immunohistochemistry (IHC) |
1:200 | - |
Immunocytochemistry (ICC/IF) |
1:100 | - |
Flow Cytometry (Flow) |
10 µg/5x10^5 cells | - |
Product Specifications | |
---|---|
Species Reactivity |
Human, Rat |
Host/Isotype |
Rabbit / IgG |
Class |
Polyclonal |
Type |
Antibody |
Immunogen |
Peptide (C)KNANFTGPNQTSSNS, corresponding to amino acid residues 21-35 of human alpha1B -adrenoceptor |
Conjugate |
Unconjugated |
Form |
Lyophilized |
Concentration |
0.8 mg/mL |
Purification |
Antigen affinity chromatography |
Storage buffer |
PBS, pH 7.4, with 1% BSA |
Contains |
0.05% sodium azide |
Storage conditions |
-20°C |
RRID |
AB_2735449 |
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 50 µL of deionized water.
The alpha-1b adrenoceptor is an Adrenergic Receptor that causes contraction of smooth muscle cells and thereby controls vascular tone, blood pressure, and accelerates the development of atherosclerosis. Decreased alpha-1b adrenoceptor is associated with benign prostatic hyperplasia (BPH). This adrenergic receptor also acts as a proto-oncogene when transfected into cell lines where constitutive activation of the receptor induces neoplastic transformation. The alpha-1b adrenoceptor has been documented in adrenal, aorta, blood, various brain regions, eye, heart, kidney, liver, prostate, spinal cord, and spleen. ESTs have been isolated from kidney (normal and cancer) and lung carcinoma libraries.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ADA1B; Adrenergic alpha 1B- receptor; adrenergic receptor, alpha 1b; adrenergic, alpha-1B-, receptor; alpha 1B-adrenoceptor; alpha 1B-adrenoreceptor; Alpha-1B adrenergic receptor; Alpha-1B adrenoceptor; Alpha-1B adrenoreceptor; alpha-1B-adrenergic receptor
Gene Aliases: ADRA1; ADRA1B; ALPHA1BAR
UniProt ID: (Human) P35368
Entrez Gene ID: (Human) 147, (Rat) 24173
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support