    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
Thermo Fisher Scientific Logo
Search Thermo Fisher Scientific
Search All
Search button Close
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          Loading
          What type of assay do you want?
          What type of assay do you want?
          Which Copy Number product(s) are you interested in using?
          What type of siRNA do you want?
          Which miRNA product(s) are you interested in using?
          Which technology are you using?
          What Array format do you want?
          What Array format do you want?
          What type of primers are you looking for?
          What species do you want to target? (Select one or more)
          What is your target of interest?
          Enter / Upload Multiple Targets
          Select a single species to search by sequence
          Enter / Upload Multiple Sequences
          Genome Version :
          for [Hs] Human
          Select a single species to search by location
          Number     Position/Start  Position/Stop
          Based on NCBI Reference Sequence: NC_012920.1
          Based on NCBI Reference Sequence: NC_012920.1
          Add more chromosome positions to your search
          Multiple chromosome co-ordinate search limit is 12.
          Upload your file (.vcf , .bed only)
          Multiple Target Search
          Option 1: Enter Keywords
          Supported Keywords:
          Supported Search Terms:
          You may conduct a batch search using the following types of search terms:
          Assay ID: e.g, "Hs00153283_m1"
          Entrez Gene ID: e.g., "10332"
          Gene Symbol: e.g., "CALCA"
          RefSeq Accession #: e.g., "NM_005795"
          GenBank RNA: e.g., "AI219647"
          Protein ID: e.g., "Q8IWW8"
          RNAdb ID: e.g., "LIT1011"
          RNAdb Gene Symbol: e.g., "Jpx"
          MicroArray Probe ID*: e.g., "208478_s_at"
          *If you enter a microarray probe ID, your search will return the 3' most assay(s) for the corresponding gene
          Assay IDs
          Entrez Gene IDs
          Gene Symbols
          RefSeq Accession numbers
          GenBank mRNA Accession numbers
          Protein IDs
          RNAdb IDs and Gene Symbols
          Array Probe IDs
          Supported Keywords:
          siRNA IDs
          Entrez Gene IDs
          Gene Symbols
          RefSeq Accession numbers
          GenBank mRNA Accession numbers
          Protein IDs
          RNAdb IDs and Gene Symbols
          Supported Keywords:
          Gene Symbol
          RefSeq mRNA Accession
          Entrez Gene ID
          UniGene ID
          Gene Alias
          GenBank mRNA Accession
          siRNA ID
          GenBank Protein ID
          Supported Keywords:
          Assay IDs
          Gene Symbols
          RefSeq Accession numbers
          DGV Accession numbers
          Supported Keywords:
          Assay IDs
          RS #s
          Gene Symbols
          RefSeq Accession numbers
          Celera hCV numbers
          Supported Keywords:
          Assay IDs
          Assay Names
          Target Sequences (mature sequences)
          miRBase IDs
          miRBase Accession Numbers
          Stem-loop IDs
          Stem-loop Accession Numbers
          Supported Keywords:
          Gene symbol (e.g. KRAS)
          COSMIC ID (e.g. 516)
          Assay Name (e.g. KRAS_rf)
          Assay ID (e.g. Hs00000174_rf)
          Nucleotide Mutation (c.2582T>A)
          Amino Acid Change (p.L861Q)
          Supported Keywords:
          Assay IDs
          Entrez Gene IDs
          Gene Symbols
          Gene Aliases
          COSMIC IDs
          SNP IDs
          Nucleotide Mutations (e.g. c.2582T>A)
          Amino Acid Changes (e.g. p.L861Q)
          RefSeq Accessions
          Supported Keywords:
          Gene symbol
          Assay ID
          Plate SKU
          Supported Keywords:
          miRBase ID
          Assay ID
          Plate SKU
          Supported Keywords:
          Gene Name
          Gene ID
          Assay ID
          Supported Keywords:
          sgRNA IDs
          Entrez Gene IDs
          Gene Symbols
          Gene Name
          Assay IDs
          Entrez Gene IDs
          Gene Symbols
          RefSeq Accession numbers
          GenBank mRNA Accession numbers
          Protein IDs
          Supported Keywords:
          Assay IDs
          Assay Names
          Target Sequences (mature sequences)
          miRBase IDs
          miRBase Accession Numbers
          Stem-loop IDs
          Stem-loop Accession Numbers
          OR
          Option 2: Upload Keywords
          The following file types are supported: ANSI files with extension .txt, .txtN (where N is a number) or .vcf containing either a list of keywords, the exported results from a previous assay search, or VCF-compliant data.
          Multiple Sequences Search
          You must enter at least one sequence before continuing
          Sequence Name: Your sequence names must be unique.
          Sequence Error: Sequences must be Entered and composed solely of the nucleotides A, C, G, T.
          Sequence Error: Sequences must be Entered and composed solely of the nucleotides A, C, G, and U.
          Sequence Name must Be Entered.
          You must enter at least one sequence before continuing
          Sequence Name: Your sequence names must be unique.
          Sequence Error: Sequences must be Entered and composed solely of the nucleotides A, C, G, T.
          Sequence Error: Sequences must be Entered and composed solely of the nucleotides A, C, G, and U.
          Sequence Name must Be Entered.
          You must enter at least one sequence before continuing
          Sequence Name: Your sequence names must be unique.
          Sequence Error: Sequences must be Entered and composed solely of the nucleotides A, C, G, T.
          Sequence Error: Sequences must be Entered and composed solely of the nucleotides A, C, G, and U.
          Sequence Name must Be Entered.
          Name
          Name
          The Sequence name must be Entered, upto 30 characters in length.
          Sequence
          Sequence:
          Please enter your sequence in the 5' to 3' direction.
          Sequences must be composed solely of the nucleotides A, C, G, T.
          Please convert U's to T's and remove any spaces,
          tabs, or line breaks.
          Sequence:
          Sequences must be composed solely of the nucleotides A, C, G, and U.
          X   Remove All
          X  Remove
          Invalid
          Valid
          X  Remove
          Invalid
          Valid
          X  Remove
          Invalid
          Valid
          + Enter More Sequences
          OR
          Option 2: Upload Sequences
          File types that are supported include FASTA files with extension .txt
          Please Upload Valid Text File.
          Error - Invalid Header in Filebuilder file. .
          Recent Searches
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States
          Your items have has been added!
          Host server : magellan-search-green-56756dfb8b-mfpfg:80/100.66.79.199:80.
          git-commit: de6905f249d25e22729e613aef254bb544bd8b7c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.1-2025.06.25-1.0