Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:1,000 | View 4 publications 4 publications |
ELISA (ELISA) |
- | View 1 publication 1 publication |
Immunoprecipitation (IP) |
2 µg/mL | View 1 publication 1 publication |
ChIP assay (ChIP) |
- | View 2 publications 2 publications |
Product Specifications | |
---|---|
Species Reactivity |
Chicken, Human, Mouse |
Published species |
Cat, Human, Yeast |
Host/Isotype |
Mouse / IgG2a, kappa |
Class |
Monoclonal |
Type |
Antibody |
Clone |
9 E11 |
Immunogen |
A synthetic peptide, corresponding to amino acid residues 408-439 (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) from the C terminus of c-myc,coupled to KLH. |
Conjugate |
Unconjugated |
Form |
Liquid |
Concentration |
0.2 mg/mL |
Purification |
Protein A |
Storage buffer |
PBS, pH 7.4, with 0.2% BSA |
Contains |
0.09% sodium azide |
Storage conditions |
4° C |
RRID |
AB_2536302 |
The c-Myc protein is a 62 kDa transcription factor that is encoded by the c-Myc gene on human chromosome 8q24. c-Myc is commonly activated in a variety of tumor cells and plays an important role in cellular proliferation, differentiation, apoptosis and cell cycle progression. The phosphorylation of c-Myc has been investigated and previous studies have suggested a functional association between phosphorylation at Thr58/Ser62 by glycogen synthase kinase 3, cyclin dependent kinase, ERK2 and C-Jun N terminal Kinase (JNK) in cell proliferation and cell cycle regulation. Studies have shown that c-Myc is essential for vasculogenesis and angiogenesis in tumor development, which distributes blood throughout the cells. The c-myc oncogene (p62 c-myc) is involved in the control of normal cellular proliferation and differentiation, and the deregulated expression of c-Myc induces apoptosis in different cell types. Antibodies against c-myc epitopes recognize overexpressed proteins containing Myc epitope tag fused to either amino- or carboxy-termini of targeted proteins. c-Myc-, N-Myc- and L-Myc-encoded proteins function in cell proliferation, differentiation and neoplastic disease. Amplification of the c-Myc gene has been found in several types of human tumors including lung, breast and colon carcinomas, while the N-Myc gene has been found amplified in neuroblastomas. Translocation of the c-myc locus on chromosome 8 to the immunoglobulin loci on chromosome 14 (heavy chain); 2 (delta light chain); or 22 (light chain) is described in Burkitts lymphoma and other B-cell lymphoproliferative conditions. An aberrant expression of the c-myc gene occurs in tumors of different origins such as colorectal, gastric, gallbladder, hepatic, mammary, ovarian, endometrial, head and neck, pulmonary, prostatic, thyroidal, oral, ocular, nasopharyngeal, endocrine, as well as hematopoietic neoplasms.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: avian myelocytomatosis viral oncogene homolog; bHLHe39; c Myc; c-Myc; c-myc epitope tag; c-myc proto-oncogene; Class E basic helix-loop-helix protein 39; cmyc; Myc proto-oncogene protein; MYC proto-oncogene, bHLH transcription factor; myc-related translation/localization regulatory factor; Proto-oncogene c-Myc; Transcription factor p64; v-myc myelocytomatosis viral oncogene homolog; v-myc myelocytomatosis viral oncogene-like protein
Gene Aliases: AU016757; BHLHE39; c-Myc; MRTL; MYC; Myc2; MYCC; Niard; Nird
UniProt ID: (Chicken) P01109, (Human) P01106, (Mouse) P01108
Entrez Gene ID: (Chicken) 420332, (Human) 4609, (Mouse) 17869
Molecular Function:
basic helix-loop-helix transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support