Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Bovine and Orangutan
eIF3D is a protein belonging to the largest of the eukaryotic translation initiation factor family, eukaryotic translation initiation factor-3 (eIF3). eIF3 is a multiprotein complex composed of at least 13 different subunits. eIF3D binds to the 40S ribosomal subunit and thus helps in maintaining the 40S and 60S ribosomal subunits in a dissociated state. An important role of eIF3D is the formation of the 40S initiation complex by interacting with the ternary complex of eIF2/GTP/methionyl-tRNA, and promotes mRNA binding. eIF3D is the major RNA binding subunit of the eIF3 complex. It associates with the subunit p170 of eIF-3. Ubiquitous expression of eIF3D is seen in most of the tissues and it is phosphorylated upon DNA damage, likely by ATM or ATR.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: eIF-3-zeta; eIF3 p66; EIF3D; Eukaryotic translation initiation factor 3 subunit 7; Eukaryotic translation initiation factor 3 subunit D; eukaryotic translation initiation factor 3, subunit 7 zeta, 66/67kDa; eukaryotic translation initiation factor 3, subunit D; RP5-1119A7.12-003; translation initiation factor eIF3 p66 subunit
Gene Aliases: EIF3D; EIF3S7
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support