Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody can label the centrosome structure.
Immunogen sequence: NWASGFSQG EKIHEDIFDI IDREADGSDS LEGFVLCHSI AGGTGSGLGS YLLERLNDRY PKKLVQTYSV FPNQDEMSDV VVQPYNSLLT LKRLTQNADC VVVLDNTALN RIATDRLHIQ NPSFSQINQL VSTIMSASTT TLRYPGYMNN DLIGLIASLI PTPRLHFLMT GYTPLTTDQS VASVRKTTVL DVMRRLLQPK NVMVSTGRDR QTNHCYIAIL NIIQGEVDPT QVHKSLQRIR ERKLANFIPW GPASIQVALS RKSPYLPSAH RVSGLMMANH TSISSLFERT CRQYDKLRKR EAFLEQFRKE DMFKDNFDEM DTSREIVQQL IDEYHAATRP DYISWGTQEQ (103-451 aa encoded by BC000619)
This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: centrosome marker; gamma tubulin; gamma-1-tubulin; gamma-2-tubulin; gamma-tubulin complex component 1; GCP-1; TUBG1; tubulin gamma 2 chain; tubulin, gamma polypeptide; unnamed protein product
Gene Aliases: 1500010O08Rik; CDCBM4; GCP-1; TUBG; TUBGCP1; Tubgl
UniProt ID: (Human) P23258, (Dog) Q9GKK5, (Human) Q9NRH3, (Rat) P83888, (Mouse) P83887, (Mouse) Q8VCK3
Entrez Gene ID: (Human) 7283, (Dog) 403695, (Human) 27175, (Rat) 252921, (Mouse) 103733, (Rat) 680991, (Mouse) 103768
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support