Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: centrosome marker; gamma tubulin; Gamma-1-tubulin; Gamma-2-tubulin; Gamma-tubulin complex component 1; GCP-1; Tubulin gamma-1 chain; Tubulin gamma-2 chain; tubulin, gamma 1; tubulin, gamma 2; tubulin, gamma polypeptide
Gene Aliases: 1500010O08Rik; AI451582; AI503389; AI504772; CDCBM4; GCP-1; TUBG; TUBG1; TUBG2; TUBGCP1; Tubgl
UniProt ID: (Human) P23258, (Dog) Q9GKK5, (Human) Q9NRH3, (Mouse) P83887, (Rat) P83888, (Mouse) Q8VCK3
Entrez Gene ID: (Human) 7283, (Dog) 403695, (Human) 27175, (Mouse) 103733, (Rat) 252921, (Rat) 680991, (Mouse) 103768
Molecular Function: tubulin
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support