Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • hnRNP A2B1 Antibodies

          Invitrogen

          hnRNP A2B1 Recombinant Rabbit Monoclonal Antibody (3B7Q8)

          View all (21) hnRNP A2B1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite hnRNP A2B1 Recombinant Rabbit Monoclonal Antibody (3B7Q8)

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (11)
          hnRNP A2B1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          hnRNP A2B1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 11

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          hnRNP A2B1 Antibody (MA569622) in ICC/IF

          Immunocytochemical analysis of hnRNP A2B1 in NIH/3T3 cells. Samples were incubated with hnRNP A2B1 recombinant monoclonal antibody (Product # MA569622) using a dilution of 1:200 (40x lens). Followed by a further incubation with Cy3 Goat Anti-Rabbit IgG (H+L) using a dilution of 1:500 (Red). The cells were counterstained with alpha-Tubulin Mouse mAb using dilution of 1:400. Followed by incubation with 488-co... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          hnRNP A2B1 Antibody in Immunocytochemistry (ICC/IF)
          hnRNP A2B1 Antibody in Immunocytochemistry (ICC/IF)
          hnRNP A2B1 Antibody in Immunocytochemistry (ICC/IF)
          hnRNP A2B1 Antibody in Immunocytochemistry (ICC/IF)
          hnRNP A2B1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          hnRNP A2B1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          hnRNP A2B1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          hnRNP A2B1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          hnRNP A2B1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          hnRNP A2B1 Antibody in Western Blot (WB)
          hnRNP A2B1 Antibody in Western Blot (WB)

          Product Details

          MA569622

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:13,000-1:78,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:3,000-1:30,000
          -

          Immunocytochemistry (ICC/IF)

          1:200-1:2,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          3B7Q8

          Immunogen

          Synthetic peptide.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1.37 mg/mL

          Amount

          137 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.3., with 50% glycerol

          Contains

          0.09% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          This gene belongs to the A/B subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has two repeats of quasi-RRM domains that bind to RNAs. This gene has been described to generate two alternatively spliced transcript variants which encode different isoforms. [provided by RefSeq, Jul 2008]

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: DKFZp779B0244; epididymis secretory sperm binding protein; FLJ22720; heterogeneous nuclear ribonucleoprotein A2; heterogeneous nuclear ribonucleoprotein B0a; heterogeneous nuclear ribonucleoprotein B0b; heterogeneous nuclear ribonucleoprotein B1; Heterogeneous nuclear ribonucleoproteins A2/B1; heterogeneous nuclear ribonucleoproteins A2/B1 {ECO:0000312|EMBL:BAF79676.1}; heterogenous nuclear ribonucleoprotein A2/B1; hnRNP A2 / hnRNP B1; hnRNP A2 protein; hnRNP A2/B1; hnRNP B1 protein; hnrnpa2b1 {ECO:0000250|UniProtKB:P22626}; HNRNPA2B1/MYC fusion; nuclear ribonucleoprotein particle A2 protein; OTTHUMP00000202465; OTTHUMP00000202466; unnamed protein product

          View more View less

          Gene Aliases: 9130414A06Rik; Hnrnp; hnrnp-A; HNRNPA2; HNRNPA2B1; HNRNPB1; HNRPA2; HNRPA2B1; HNRPB1; IBMPFD2; OPMD2; RNPA2; SNRPB1

          View more View less

          UniProt ID: (Human) P22626, (Rat) A7VJC3, (Mouse) O88569

          View more View less

          Entrez Gene ID: (Human) 3181, (Rat) 362361, (Mouse) 53379

          View more View less

          Function(s)
          nucleic acid binding RNA binding mRNA 3'-UTR binding protein binding miRNA binding identical protein binding single-stranded telomeric DNA binding DNA polymerase binding G-rich strand telomeric DNA binding molecular condensate scaffold activity N6-methyladenosine-containing RNA binding nucleotide binding regulatory region RNA binding poly(A) RNA binding pre-mRNA intronic binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter mRNA processing RNA splicing male gonad development lung development negative regulation of mRNA splicing, via spliceosome oligodendrocyte differentiation RNA transport response to mineralocorticoid mRNA splicing, via spliceosome mRNA export from nucleus primary miRNA processing DNA geometric change mRNA transport regulation of macromolecule metabolic process positive regulation of telomere maintenance via telomere lengthening miRNA transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-6479c7f679-sz4vp:80/100.66.76.236:80.
          git-commit: b70c0dceb888abc3b0dcebb5bc3a5b8753e7e816
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.50.0-Offline