OriGene
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
Assay-Dependent | - |
Product Specifications | |
---|---|
Species Reactivity |
Tag |
Host/Isotype |
Mouse / IgG2b |
Class |
Monoclonal |
Type |
Antibody |
Clone |
OTI4E10 |
Immunogen |
Recombinant protein of mOrange expressed in HEK293 cells. It recognizes both mOrange and mOrange2 proteins. |
Conjugate |
Unconjugated
|
Form |
liquid |
Concentration |
3.35 mg/mL |
Purification |
Protein G |
Storage buffer |
PBS with 1% BSA, 50% glycerol |
Contains |
0.02% sodium azide |
Storage conditions |
-20° C, Avoid Freeze/Thaw Cycles |
mOrange/mOrange2 fluorescent protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mOrange
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support