Search
Discrete industrial Analyzers (DIA) provide rapid and automated photometric (Colorimetric and enzymatic) and electrochemical (pH and conductivity) analysis giving fast, reproducible results. With discrete cell technology, multiple analytes can be run simultaneously reducing operation and run time.
This webinar will cover how Discrete Industrial Analyzers work and the Thermo Scientific™ Gallery™ and Gallery™ Plus systems, with automation enabling up to 350 tests per hour. Our DIA Specialist will also discuss how this technique is an efficient alternative to traditional flow analyzers and how the unique low volume cuvette design produces cost savings in reagents and waste. Finally we show how this technology can be applied in an environmental laboratory for tests such as alkalinity, conductivity, nitrite, urea, total oxidized nitrogen (TON), silica and total hardness in a variety of sea, natural and waste waters and soil testing.
Key learning objectives:
Who should attend?
James Hannan
Application Specialist Discrete Industrial Analyzers | Thermo Fisher Scientific, Hemel Hempstead, UK
*Required fields