On this page:
Consistent bioprocess control designed for scalability
The Thermo Scientific HyPerforma G3 family of bioprocess controllers offers flexible and reliable advanced control capabilities. The controllers are fully self-contained, movable units that can be operated alone (for one vessel) or networked for multiple vessels. They are engineered to optimize capital cost for use in non-GMP and CGMP-certified production facilities. Together, the control and automation platform scales from bench through production. Integrating flexibility and an open-architecture design, this platform allows the use of third-party vessels, as well as Thermo Scientific single-use bioprocessing equipment.
Find a controller that fits your needs
Select a workflow step to see which HyPerforma G3 Bioprocess Controller to use:
Step 1: Drug discovery and R&D
Thermo Scientific HyPerforma G3Lab Bioprocess Controller
Easy process scale-up or scale-down
The G3Lab Controller can control most brands of single-use or autoclavable bioreactors and fermentors that are <50 L, including stirred-tank and rocking models. The enclosure contains state-of-the-art transmitters along with power supplies, pumps, input/output (I/O) modules, and the hardware required to connect to the control network, providing maximum control capability.
Key features:
  • Scalability to <50 L
  • Open-architecture capabilities to integrate with vessels from other suppliers, such as the Eppendorf BioBLU Single-Use Vessels and Sartorius UniVessel Benchtop Bioreactors
  • The ability to build and manage complex, multi-feed dosing strategies
  • Allows for third-party peripheral integration as needed
  • Touchscreen interface for easy data entry and control
  • Redundant pH and DO Sensors
  • Customizable gas flow control
  • Verified and validated with TruBio software
Step 2: Process development
Thermo Scientific HyPerforma G3Lite Bioprocess Controller
Modularity for scale-up
The G3Lite Bioprocess Controller is an open-architecture control system that can be integrated with most bioreactors and fermentors—regardless of the manufacturer. The system consists of a control tower that leverages intelligent transmitters, mass flow controllers (MFCs), pumps, and sensor controls. This controller is fully self-contained, mobile, and can be operated as stand-alone (for one vessel) or networked for multiple vessels. They are engineered to optimize capital cost for use in non-GMP and cGMP-certified production facilities.
Key features:
  • Scalability: Volumes 50 L to 2,000 L
  • Modularity: Predefined configurations available for 50, 100, 250, 500, 1,000 and 2,000 L bioreactors
  • Touchscreen interface for easy data entry and control
  • Stand-alone or networked (for multiple vessels)
  • Redundant pH and DO sensors
  • Verified and validated with TruBio software
Step 3: Clinical manufacturing
Thermo Scientific HyPerforma G3Pro Bioprocess Controller
Versatility for scale-up
The G3Pro Bioprocess Controller offers many of the G3Lite Bioprocess Controller capabilities as an open-architecture, universal controller used to control most bioreactors and fermentors. These controllers enable an additional layer of versatility, allowing mobility and flexibility in terms of providing options for any reconfiguration or application expansion. They are engineered to optimize capital cost for use in non-GMP and cGMP-certified production facilities.
Key features:
  • Scalability: Volumes 50 L to 5,000 L
  • Open-architecture capabilities to integrate with vessels from other suppliers
  • Single or dual cart mount option for easy mobility and reduced footprint
  • Touchscreen NEMA interface for easy data entry and control
  • Probe configuration options: Flexible to connect electrochemical, single-use, or both for pH and DO supports variable cell density measurement
  • Redundant sensor control mechanism
  • Can be adapted for multi-product applications
  • Supports variable cell density measurement
  • Verified and validated with TruBio software
Supporting products used across the workflow from R&D to manufacturing
TruBio Bioprocess Automation Software
The TruBio software, powered by the Emerson DeltaV™ Distributed Control platform, offers easy-to-configure process control so learning automation control programming is not necessary. Developed for use with all Thermo Scientific controllers, TruBio software is designed to support easy scaling, data integrity and technology transfer and allow the ability to build sophisticated process control strategies. It offers flexibility to incorporate a wide range of equipment and manage multiple data streams from several unit operations. TruBio software has been developed according to GAMP 5 methods and conforms to regulatory requirements for use in cGMP-compliant processes.
Bioreactor systems from lab to commercial manufacturing
Discover the advantages of a comprehensive single-use bioprocessing equipment portfolio, enabling scalability from research to cGMP manufacturing that will allow you to choose the ideal match for your process parameters.
FlexCare Services
We understand that reliable equipment is crucial to your bioprocessing operations. That’s why we offer FlexCare services to help your single-use equipment operate at its best while helping you maximize productivity. Whether you’re focused on innovation and speed or optimizing your workflow, FlexCare services can empower your team to stay focused on your important work. We have a selection of service plans—from bench-scale to manufacturing, designed to meet your needs.
FlexCare Services
For Research Use Only. Not for use in diagnostic procedures.