Learn more and explore available resources for the Genexus Dx Purification System and NGS research
Downloadable guides
Brochure: The Genexus Dx System
Genexus Dx brochure cover
Instrument services and support guide
Instrument services and support guide brochure cover
Intended Use: The Genexus Dx Purification System is a sample preparation instrument which automates nucleic acid extraction, purification, and quantification of DNA and RNA from FFPE tissue sections for use in next-generation sequencing applications. The system uses MagMAX magnetic bead-based technology to perform nucleic acid extraction. Purified nucleic acid is quantified using the onboard Qubit fluorometer which uses fluorescence to measure nucleic acid.
For In Vitro Diagnostic Use.
CN: 63521