The Thermo Scientific Celestron transmission line pulse/very fast transmission line pulse (TLP/VF-TLP) Test System can be configured for both Standard TLP and VF-TLP for testing at the wafer and/or package level. Optional probes can be used to measure signals on pins or pads other than the ones being stressed. The Celestron system software is the most comprehensive in the industry and utilizes graphics to assist in system setup and connection to the device under test (DUT). During test operations, it displays the recorded TLP pulse voltage and current waveforms, compiled pulsed I-V curve, leakage current measurements, and DC I-V curve trace data. The operator can select the range of test voltages (stress pulses), pulse polarity, leakage and curve trace parameters. The position and duration of the measurement window within the TLP pulse can also be selected, and modified after the data is collected.
Key Features
Flexible test capabilities
The Thermo Scientific Celestron TLP/VF-TLP Test System can be configured for Standard TLP and VF-TLP for testing at the wafer level and/or package level. Optional probes can also be used to measure signals on pins or pads other than the ones being stressed. 
Unsurpassed test control
The operator can select the range of test voltages (stress pulses), pulse polarity, leakage and curve trace parameters. The position and duration of the measurement window within the TLP pulse can also be selected, and modified after the data is collected.
Pulse generator
High current TLP pulse generator.
Probers
Can be interfaced with semiautomatic probers.
Specifications
Style Sheet for Products Table Specifications
Standard TLP configurationPulse widths from 30 to 500ns
Integrated in the pulser box, the standard TLP Pulse width is 100ns
Additional pulse widths can be added externally through a cable change ranging from 30 to 500ns
Optional computer controlled pulse width with 3 selectable pulse widths
Rise times available
Standard rise time is 200ps
Optional rise times from 500ps to 10ns
Controlled by optional external rise time filters
2ns and 10ns external rise time filter included
Optional computer controlled rise time with 3 selectable rise times
Note: 500 MHz or faster oscilloscope is required for TLP measurements
1 GHz or grater scope is recommended for measurements with rise times of less than 1ns
Support of all TLP configurations
Optional changing of configuration for 25 to 500 ohm delivery impedances
Support for all configurations described in the ESD Association’s TLP Standard Test Method
Both wafer level and package testing at all impedances
Standard TLP / VF-TLP configurationIntegrated system controller
No need for a dedicated computer
Windows® operating system
Integrated source/meter unit
Curve Tracing to ± 200V
Leakage measurements down to 50pA
DUT failure detection based on leakage or voltage with forced current or both
Transmission Line Pulse
(TLP / VF-TLP)
Designed in compliance with the current ESD Association Standard Test Method Documents (ANSI/ESD STM 5.5.1-2016)
Standard VF-TLP configurationPulse widths from 1.2 to 10ns
Selected by cable change at front of system
Standard widths are 1.2, 2.5 and 5ns (others optional)
Rise times from 200ps to 2ns
Controlled by optional filters
Standard rise times are 200 and 300ps
2ns external rise time fi ler included
Maximum pulse current
15A into a 50 ohm load
Approximately 30A into a short circuit
Maximum open circuit voltage
1500V
Applications
esd_thumb_274x180_144dpi
Cualificación de semiconductor ESD
Se necesita un plan de control de descarga electrostática (ESD) para identificar los dispositivos que son sensibles a ESD. Ofrecemos un conjunto completo de sistemas de prueba para ayudarle con los requisitos de cualificación de su dispositivo.
Style Sheet for Komodo Tabs
Techniques
Pruebas de conformidad con ESD
La descarga electrostática (ESD) puede dañar pequeñas características y estructuras en semiconductores y circuitos integrados. Ofrecemos un completo conjunto de equipos de prueba que verifica que sus dispositivos cumplen con los estándares de conformidad ESD.
Más información ›
Pruebas de conformidad con ESD
La descarga electrostática (ESD) puede dañar pequeñas características y estructuras en semiconductores y circuitos integrados. Ofrecemos un completo conjunto de equipos de prueba que verifica que sus dispositivos cumplen con los estándares de conformidad ESD.
Más información ›
Style Sheet to change H2 style to p with em-h2-header class
Contact us
Electron microscopy services for
semiconductors
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
Learn more ›