3-images-670x119_Rv2
Isohumulones (iso-α-acids) form approximately 80%of the typical bitterness of beer. Their antimicrobial effect leads to a sterile beverage; their tensioactive character stabilizes the foam; and they have a major influence on the general flavor, smell, and smoothness of beer. HPLC represents the only analytical method that enables the specific quantitation of iso-α-acids in beer.
Thermo Scientific™ BeerNHop Solution provides a complete determination and quantitation of all major cis- and trans-isomer variants of iso-α-acids that are present.
The solution consists of a Thermo Scientific™ Dionex™ UltiMate™ 3000 system package and its related Thermo Scientific™ Isohumulones Starter Kit. System packages contain Thermo Scientific™ Dionex™ Chromeleon™ 7.2 Chromatography Data System software, as well as the Dionex UltiMate 3000 hardware needed to set up an entire UHPLC+-focused system for measuring the isohumulones. The starter kit provides all of the consumables, thereby the analysis can be started immediately. For more information, please download the BeerNHop Solution brochure or contact your local representative.
Downloads:
Thermo Scientific Dionex UltiMate 3000 System Packages
Complete UltiMate 3000 System descriptions
Part number
Thermo Scientific Dionex UltiMate 3000 RS System Package with On-Line SPE
5200.0500
Thermo Scientific Dionex UltiMate 3000 SD System Package with On-Line SPE
5200.0505
Thermo Scientific Dionex UltiMate 3000 SD System Package with Direct Injection
5200.0510
Complete Starter Kit descriptions
Part number
Thermo Scientific Isohumulones Starter Kit for On-Line SPE RS System
TS-MKIT0012
Thermo Scientific Isohumulones Starter Kit for On-Line SPE SD System
TS-MKIT0013
Thermo Scientific Isohumulones Starter Kit for Direct Injection SD System
TS-MKIT0014
Individual Items descriptions
Part number
Thermo Scientific™ Pierce™ Acetonitrile, 1 L
TS-51101
Thermo Scientific™ Pierce Water, 1 L
TS-51140
Thermo Scientific™ Formic Acid, 10 mL
TS-0001010
Thermo Scientific™ Ethylenediaminetetraacetic acid, 100 mg
TS-0002004
Thermo Scientific™ Isohumulones standard, pack of 5 × 2 mL ampoule
TS-7004002
Thermo Scientific™ Hypersil GOLD™, 1.9 μm, 2.1 × 100 mm
25003-153030
Thermo Scientific Hypersil GOLD, 3 μm, 3 × 150 mm
25003-153030
Thermo Scientific Hypersil GOLD C8, 5 μm, 2.1 × 20 mm
25205-022130
Thermo Scientific™ Humulones and Lupulones standard, pack of 5 × 2 mL ampoule
TS-7003002
Thermo Scientific™ Tetrahydroisohumulones standard, pack of 5 × 2 mL ampoule
TS-7005002
Disposable Syringes,1 3mL, luer lock, pack of 100 pcs.
S7515-3
Thermo Scientific™ Target™2 Syringe Filter,1 GMF, 0.7 μm, pack of 100 pcs
F2500-18
 1 Recommended to be used for wort analyses
How to order
USA and Canada
800 332 3331
865 354 4616 fax
Email
France
+33 (0)1 60 92 48 34
+33 (0)1 60 92 49 00 fax
Email
Germany
+49 (0) 2423 9431 -20 or -21
+49 (0) 6103 408 1640 fax
Email
United Kingdom
+44 1928 534110
+44 1928 534001 fax
Email
India
+91-22-6742 9494
+91-22-6742 9495 fax
Email
Japan
+81 3 5826 1615
+81 3 5826 1611 fax
Email
China
800-810-5118
Shanghai: +86-21-68654588
Bejing: +86-10-84193588
Guangzhou: +86-20-83145199
Email
Thermo Fisher Scientific
Australia Pty Ltd
1300 735 292 (free call domestic)
1800 067 639 fax
Email
Thermo Fisher Scientific
New Zealand Ltd
0800 933 966 (free call domestic)
0800 329 246 fax
Email
All Other Enquiries
+44 (0) 1928 534 050
+44 (0) 1928 534 049 fax
Email
Support
North America
800 332 3331
Email
Outside North America
+44 (0) 1928 534 440
Email