Glacios TEM
Transmission Electron Microscopes (TEMs) fire a beam of electrons through an ultra-thin sample specimen. The electrons interact with the sample as they pass through it, producing information about the internal structure of the sample. Many of our TEMs feature fully automated operation and are capable of sub-Ångström resolution.
Popular products
Glacios Cryo-TEM
Talos F200X TEM
Themis S S/TEM
Glacios Cryo-TEM
The new Thermo Scientific Glacios Cryo Transmission Electron Microscope (Cryo-TEM) delivers a complete and affordable Cryo-EM solution to a broad range of scientists. It features 200 kV XFEG optics, the industry-leading Autoloader (cryogenic sample manipulation robot) and the same innovative automation for ease of use as on the Krios G3i Cryo-TEM. The Glacios Cryo-TEM bundles all this into a small footprint that simplifies installation.
Available for: Life Sciences Research
Themis TEM
Themis TEM
Themis transmission electron microscope (TEM) starts with our revolutionary wide high-tension range and corrector-ready platform, combined with a wide pole piece gap delivering the space to do more. Developed with stability and flexibility in mind, the Themis TEM high performance continuously enables atomic scale discovery and exploration. 
Product Models:
Themis S S/TEM for Semiconductors
Themis Z S/TEM for Materials Science
Themis ETEM for Materials Science
Available for:  Materials Science and Semiconductor Research
Metrios TEM
Metrios transmission electron microscope (TEM) is the first TEM dedicated to providing the fast, precise measurements that semiconductor manufacturers need to develop and control their wafer fabrication processes.
Available for: Semiconductors Research
Krios G3i Cryo-TEM
The new Thermo Scientific Krios G3i Cryo Transmission Electron Microscope (Cryo-TEM) enables life science researchers to unravel life at the molecular level—easier, faster, and more reliably than ever before. Its highly stable 300 kV TEM platform and industry-leading Autoloader (cryogenic sample manipulation robot) are designed for automated applications, such as single particle analysis (SPA) and cryo-tomography. Designed-in connectivity ensures a robust and risk-free pathway throughout the entire workflow, from sample preparation and optimization to image acquisition and data processing.
Available for: Semiconductors Research
Talos TEM
Talos TEM
Talos transmission electron microscope (TEM) is built to deliver rapid access to 2D and 3D data so that you can concentrate on discovery. With configurations for materials research and life sciences research, the Talos TEM is a truly versatile system with many innovations that will serve your research needs for years to come.
Product Models:
Talos F200i for Materials Science
Talos L120C TEM for Materials Science
Talos L120C TEM for Life Sciences
Talos F200X TEM for Materials Science
Talos F200S TEM for Materials Science
Talos Arctica TEM for Life Sciences
Talos F200C TEM for Life Sciences