Search
Our advanced technologies are designed to support the global transition to clean energy. From renewable energy production to hydrogen generation and carbon capture, our solutions help drive sustainability and efficiency. Whether in research and development or large-scale production, our technologies help to ensure every process is optimized for performance and environmental responsibility.
e-fuels are synthesized from renewable hydrogen and captured CO2, offering a sustainable, carbon-neutral alternative to traditional fossil fuels. These fuels can be integrated into existing infrastructure and used in conventional combustion systems, making them a versatile and immediate solution for reducing carbon emissions.
Biofuels, derived from organic materials such as plants, algae, and waste, are a renewable energy solution that reduces dependency on crude oil. They contribute to waste reduction while providing a cleaner, more sustainable alternative to fossil fuels.
Syngas, produced from biomass or other organic materials, is a flexible fuel that can be used for electricity generation or as a building block for creating other fuels and chemicals. It is an essential part of diversifying energy sources and moving towards cleaner alternatives.
Hydrogen is one of the cleanest fuels available, and its production is pivotal to the future of energy. The production of hydrogen must be precise to ensure that it is of the highest purity and efficiency, which is where our technology comes into play.
Real-time monitoring with Raman spectroscopy can be used as a process measurement technique for hydrogen production. The Thermo Scientific MarqMetrix All-In-One Process Raman Analyzer, paired with the Thermo Scientific MarqMetrix FlowCell Sampling Optic, provides in-line, non-destructive analysis of hydrogen spin isomers, purity, phase transitions, temperature, and pressure effects, providing insights into the efficiency and purity of hydrogen production methods.
Carbon capture is essential to reducing carbon emissions and mitigating climate change. Our technologies support this process by enabling accurate measurement, monitoring, and reporting of CO₂ throughout the capture and storage lifecycle. For storage and sequestration, systems calculate, store, monitor, and transmit data on the volumetric or mass flow being securely managed—ensuring efficient, compliant, and environmentally safe carbon handling.
For Research Use Only. Not for use in diagnostic procedures.