Nuclear radiation detection and monitoring Our nuclear radiation detection
Optimize safety, operational efficiency, and regulatory compliance
Our instruments and services are designed to deliver the latest advancements in technology and functionality and help you sustain safety and productivity long into the future.
  • Breadth—full product suite for a wide range of nuclear radiation detection needs
  • Quality—designed and manufactured for a lifetime of use in the harshest of environments
  • Experience—serving the industry for over 55 years
  • Innovation—new technologies and solutions to help ensure you keep pace with a changing industry
Continuous area monitoring to optimize efficiency and worker safety
The Thermo Scientific™ RadHalo™ RDP and FM Spectoscopic Area Monitors detect and identify gamma radiation in the environment. With configurations for fixed-site perimeter monitoring as well as mobile deployment, RadHalo area monitors ensure worker safety by providing high-confidence nuclear radiation measurements when and where you need them.
  • Reduce costs while receiving timely and detailed radionuclide information 
  • Replace labor-intensive environmental instruments with an automated system 
  • Early warning system through continual dose-rate monitoring
Download the RadHalo product brochure
Download the RadHalo RDP and FM specification sheet
Video
Watch the RadHalo monitor at work
Video
Take a look inside our facilities.
Handheld detection for any scenario
Thermo Scientific™ RadEye™ Spectroscopic Radiation Detectors are ideal for contamination monitoring in radiation controlled areas, contamination check points, and waste and material shipping surveys.
  • Portable—pager-sized detectors are easy to carry and go with you wherever nuclear radiation detection and monitoring is needed
  • Configurable—units can be customized to match a wide variety of applications
  • Easy-to-use—highly intuitive user interface helps ensure rapid adoption and wide-spread use
Download the RadEye product selection guide
Overview of RadEye Spectroscopic Radiation Detectors for nuclear power facilities
Current challenge Model to solve Benefits
Heavy meters for dose for monitoring ambient dose rate RadEye G Gamma dose rate readings in a pager-sized instrument
Time-consuming sample gathering and lab-based counting needed to identify isotope present in survey result RadEye SPRD Immediately identifies isotope present so you can act quickly
Multiple devices needed to survey nuclear radiation waste and shipping materials Range of RadEye instruments One lightweight device can measure multiple radiation types
Large and heavy Rem Ball and meter are difficult to manage for neutron surveys RadEye NL Less than 8 lbs
Emergency planning requires multiple large inventory instruments RadEye SX,
RadEye G
and RadEye B20 ER
Fewer, small handheld instruments minimize prep and ownership costs
Field contamination alpha/beta counting takes a long time to complete RadEye HEC Delivers counting room accuracy in a handheld device that can be used anywhere
Upgrade costs for new probes when changing instruments RadEye X Series Modern, digital meter can be used with most existing probes
Additional services and information for nuclear power facilities
Nuclear power plant outage services
Take advantage of configurable pre-outage, outage, and post-outage support programs offered by our certified factory trained experts to maximize your nuclear power plant efficiency, comply with critical timelines, and manage unplanned costs.
Download the outage services brochure
With more than 55 years of experience providing products and services to the nuclear power industry, we provide the experience to help you meet your business goals of plant reliability and equipment uptime. From installation and certification services to preventative maintenance, technical support, training, spare parts, and global support capabilities, we provide a wide variety of integrated solutions and instrument upgrade packages to maximize your nuclear power plant performance.
Find out more about integrated services
By installing Thermo Scientific Neutron Flux Monitoring systems, you achieve peace of mind with higher reliability, lower maintenance costs and better accuracy. Our rugged, field-proven components and streamlined design deliver superior reliability and qualify it to last the life of the reactor. No longer must you be forced to routinely replace short-lived detectors or continually service obsolete electronics. We can help you upgrade and standardize your systems, consolidating your engineering and procurement, trim your vendor list, lower your total cost of ownership, and enhance your profitability.
Request more information