Cytokine, Chemokine, & Growth Factor ELISA Kits
Measure secreted, circulating proteins in biological samples
Cytokines are secreted proteins that play a role in both the induction and effector phases of all immune and inflammatory responses, including cell proliferation, intercellular communication, and cell death. Chemokines are small cytokines which attract and recruit other cells into a localized area to exert their biologic effects. Growth factors include both cytokines and protein hormones, which stimulate proliferation as well as differentiation and maturation of responsive cells.
Cytokines have been implicated not only in immunological conditions and inflammatory response, but also in disease states such as cancer and neurodegenerative disorders. The accurate quantitation of cytokines in biological samples such as plasma, serum, and tissue culture samples enables researchers to fully understand the mechanisms behind disease in physiological conditions.
Features of Invitrogen cytokine ELISA kits
Invitrogen ELISA kits are developed to meet industry-standard specifications including standard calibration, precision, sensitivity, specificity, recovery, lot-to-lot consistency, and parallelism. Each protein is researched to target physiologically relevant sensitivity and tested in well-established models when available. These kits are manufactured in an ISO facility with stringent quality control to help ensure excellent quality and reproducibility.
-
Broad menu of over 700 targets - Optimized for sensitive, accurate, and consistent performance
- Thorough instructions to complete protocol in 2.5 to 4 hours (varies by kit)
- Validated for typical sample types (e.g., serum, plasma, supernatant, lysates)
- Available as ready-to-use Coated ELISA kits and coat-it-yourself Uncoated ELISA kits
Select cytokine, chemokine, and growth factor ELISA kits from among all target-specific ELISA kits using the following selection guide.
Cytokine ELISA kit demo videos
Additional information
Learn more about cytokine and related cell signaling pathways and search for specific antibodies.
Cytokine, Chemokine, Growth Factor Pathways
Get the complete immunoassay guide to protein quantitation.
Immunoassay guide to protein quantitation
Select antibodies to build your own ELISA
We offer a variety of specific antibodies to help you.
Let us develop a custom ELISA or ProQuantum immunoassay for you
We provide development services of your chosen protein targets and immunoassay platform.
Find related products for your ELISA workflow
ELISA Reagents and Buffers ELISA Microplates and Plasticware ELISA Instruments and Equipment
Get help with your experiment from one of our scientists
Contact a field application scientist
Resources
Learning
Support
Related resources
For Research Use Only. Not for use in diagnostic procedures.