hemidesomosome
We offer one of the most extensive portfolios of antibodies for studying cell adhesion and cell junctions, including, antibodies to study all five types of cell junctions, exclusive ABfinity™ antibodies, and antibody-labeled conjugates to visualize a broad target range, with high specificity
Popular products
What are cell junctions?
Cell junctions regulate small-molecule trafficking between cells, the organization of cells into tissues, and the adherence of cells to each other and the extracellular matrix. Dysfunctional junctions are implicated in several diseases, including cancer, neurodegeneration, and cardiovascular dysfunction. Understanding cell junctions is fundamental to understanding disease pathology.
Gap junctions
Connexin1-sc
Figure 1—Connexin-40 and Connexin-43. Immunofluorescent staining of mouse heart atrium using mouse anticonnexin-43 antibody (red) and mouse anti-connexin-40 antibody (green). Image courtesy of James I. Nagy, PhD, University of Manitoba, Canada.
Gap junctions are clusters of tightly packed channels that allow small molecules (metabolites, second messengers, and ions) to travel between adjoining cells1. This molecular exchange is essential during development and differentiation; it also creates cell proliferation, tumor suppression, cell death propagation, and survival-modulating signals2. These signals include connexins, and pannexin 1 and 2, important components of synapses3.. Several kinases phosphorylate connexins and alter their functions: V-src kinase, C-src kinase, PKC, MAPK, cdc2 kinase, casein kinase I, and PKA.
A number of other proteins, such as ZO-1, ZO-2, β-catenin, drebrin, α/β tubulin, caveolin-1, NOV, CIP85, and OCP2 also associate with connexins. Connexin-43 (CX43) and connexin-40 work together to form cell junctions that are essential for proper heart atrium cell function (Figure 1). 
Tight junctions
Figure 2. Claudin proteins. Immunofluorescence staining of frozen mouse jejunum tissue using Mouse anti-Claudin-1 (red), and Mouse anti ZO-1 is labeled in green; nuclei are stained with DAPI (blue). Image courtesy of Jennifer Holmes and Dr. James Anderson, University of North Carolina at Chapel Hill, NC.
Tight junctions are barriers that regulate the paracellular movement of solutes down their electro-osmotic gradients. They also act as a fence that maintains differential composition of basolateral and apical membrane domains, limiting the diffusion of lipids and proteins between cellular compartments. The main components of tight junctions are claudins, occludins, and junctional adhesion molecules (JAMs). Proteins such as ZO-1, ZO-2, ZO-3, MUPP-1, and PATJ colocalize with claudin proteins and together form tight junctions (Figure 2). 
Claudin2-sc
Adherens junctions
junctions3-sc
Figure 3. Cell junctions. Immunohistochemistry of mouse hippocampus using AF-6 ABfinity™ Recombinant Rabbit Monoclonal Antibody, at 1 μg/mL. Goat anti Rabbit Alexa Fluor® 488 was used as secondary antibody. Image provided by Jim Nagy, Canada.
Adherens junctions are the building blocks of tissue architecture. Their main components are cadherins and catenins. These proteins form cell−cell junctions (Figure 3), and can facilitate signaling pathways such as AKT, Wnt, and MAPK to govern morphogenesis, tissue homeostasis, and intercellular communication5.. Disruption of adherens junctions promotes tumor cell invasion and metastasis 6
Desmosomes and hemidesmosomes
Desmosomes and hemidesmosomes are specialized in cell−cell and cell–extracellular matrix adhesion, respectively7. They are important in cytoskeletal organization, cell signaling, and tissue patterning. The main components of desmosomes are cadherins (desmogleins and desmocollins). Hemidesmosomes employ integrins as their primary cell adhesion proteins.
bulk-pod-showcase-270x195
Partner with us to scale-up your process or product development pipeline with bulk quantities of high-quality immunoassay reagents and biochemicals. We offer a variety of compounds, devices, and resins for your large-scale applications.  
Complete request form