Search
Developing vaccines is a complex process. Manufacturers must navigate strict regulations while also trying to predict global demand and tackle the challenges of producing large quantities of high-quality vaccines. Time-to-market and cost effectiveness are always top of mind. To speed up development and make manufacturing smoother, it's essential to find the right media, tools, and support.
We understand the challenges you face in bioprocessing, and we're here to help. With our high-performance cell culture media, tools, and knowledgeable support, we aim to make your workflow smoother, reduce risks, and enhance cost effectiveness.
| Gibco OptiPRO SFM AGT Medium | Gibco TrypLE Select Enzyme |
![]() | ![]() |
| Serum-free, animal-origin-free culture medium designed for growth of several kidney-derived cell lines, including MDBK, MDCK, PK-15, and VERO, for virus or recombinant protein production. In addition, OptiPRO SFM has been used to successfully grow other cell lines, such as Hela, BHK-21, and COS-7. | Animal origin free (AOF), recombinant enzyme for dissociating a wide range of adherent mammalian cells, including CHO, HEK 293, A529, primary human keratinocytes, and embryonic stem cells. We offer a variety of TrypLE formulations for a range of cell culture applications. |
| Format: Advanced Granulation Technology (AGT) | Format: Liquid |
|
|
| Read application note | Read application note |
| Order now | Order now |
We offer an integrated array of services to simplify cell line development through Gibco Bioproduction Services. From vector design to cGMP cell banking, our experienced scientists and project managers are able to provide total life cycle support.
Our chemically defined and serum-free media for virus and vaccine production deliver cell growth and virus production equivalent or superior to serum-supplemented systems while also maximizing consistency and reliability and simplifying downstream purification.
POROS Chromatography resins are today’s best performing chromatographic media for process-scale bioseparations. POROS resins are available in a range of selections designed especially for today’s recombinant protein and other biological purification challenges.
Applied Biosystems SEQ rapid molecular methods for pharmaceutical manufacturing help ensure the quality and safety of your pharmaceutical products especially when accuracy and time-to-results are critical. SEQ rapid molecular methods are also ideal for use in public health laboratories, cosmetics and personal care products manufacturing, food testing laboratories, and academic and research centers.
We offer an integrated array of services to simplify cell line development through Gibco Bioproduction Services. From vector design to cGMP cell banking, our experienced scientists and project managers are able to provide total life cycle support.
Our chemically defined and serum-free media for virus and vaccine production deliver cell growth and virus production equivalent or superior to serum-supplemented systems while also maximizing consistency and reliability and simplifying downstream purification.
POROS Chromatography resins are today’s best performing chromatographic media for process-scale bioseparations. POROS resins are available in a range of selections designed especially for today’s recombinant protein and other biological purification challenges.
Applied Biosystems SEQ rapid molecular methods for pharmaceutical manufacturing help ensure the quality and safety of your pharmaceutical products especially when accuracy and time-to-results are critical. SEQ rapid molecular methods are also ideal for use in public health laboratories, cosmetics and personal care products manufacturing, food testing laboratories, and academic and research centers.
Application note: Diploid Growth Serum-Reduced Medium can reduce or eliminate serum use in human and animal vaccine production
Brochure: Streamline viral vector and vaccine development
Brochure: Vaccine Production Cell Culture
Poster: Development of a chemically defined medium for Escherichia coli, Gibco Bacto CD Supreme Fermentation Production Medium (FPM) leveraging learnings from traditional peptone-based media
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing.
Explore our resources
Download Bioprocessing Product Reference Guide
For research use or further manufacturing. Not for diagnostic use or direct administration into humans or animals.