PerCP-eFluor 710 spectra excitation shown as dotted line and emission shown as solid red histogram
4488710/50482708(in buffer) 1
flow cytometry
Invitrogen PerCP-eFluor 710 is a blue laser–excitable tandem dye designed for use in flow cytometry applications. It was developed as an optimal choice for the FL3 channel on a blue laser line. It is consistently two to three times brighter than PerCP-Cyanine5.5 when evaluated in clone-to-clone comparison tests on identical samples. Although PerCP-eFluor 710 is slightly red-shifted in its emission compared to PerCP-Cyanine5.5 (710 nm vs. 685 nm), it uses the same filter sets. No change to your instrumentation is needed if you already use PE-Cy5.5 or PerCP-Cy5.5, allowing you to easily add PerCP-eFluor 710 to your multicolor panels.
We offer PerCP-eFluor 710 dye conjugated to a variety of antibodies, proteins, and amplification substrates optimized for cellular labeling and detection.
Search PerCP-eFluor 710 primary antibodies  Search PerCP-eFluor 710 secondary antibodies
  • Greater mean fluorescence intensity (MFI) as compared to PerCP-Cyanine5.5
  • Minimizes the need for compensation out of the PE detector
  • Stable performance when treated with formaldehyde fixation or exposed to ambient light
Flow histogram comparing PerCP-eFluor 710 dye to PerCP-Cyanine5.5 dye
Comparison of staining performance of PerCP-eFluor 710 to PerCP-Cyanine5.5. Mouse splenocytes were stained with CD4 Monoclonal Antibody (RM4-5), PerCP-eFluor 710 (red line) or CD4 Monoclonal Antibody (RM4-5), PerCP-Cyanine5.5 (pink line).
Flow histogram comparing PerCP-eFluor 710 dye before and after aldehyde fixation or extended ambient light exposure
Stability of PerCP-eFluor 710 dye. Human peripheral blood cells (PBMCs) were stained with CD4 Monoclonal Antibody (SK3 (SK-3)), PerCP-eFluor 710 and analyzed immediately (red line). The sample was then divided and either exposed to ambient light for six hours (blue line) or fixed in 2% formaldehyde overnight at 4°C (green line) and analyzed again. 
Cy™ is a trademark or registered trademark of GE Healthcare.
Not for resale. Super Bright Polymer Dyes are sold under license from Becton, Dickinson and Company.
Brilliant Ultra Violet™ and Brilliant Violet™ are trademarks or registered trademarks of Becton, Dickinson and Company or its affiliates, and are used under license.
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments