Enhanced-ULT-278x195
Exceptional performance for everyday assurance.
Protect your precious samples and reduce your energy costs with an ultra-low temperature freezer (ULT) that's built to last. Providing the dependability your -80°C storage requires with new energy-savings and sustainability features, our advanced ultra-low freezers feature a range of upright models, maximizing storage capacity from 30,000 up to 60,000 2mL vials. A newly enhanced touchscreen interface with capacitive touch enables easy access to set points, data and other performance and security information.
  • US EPA SNAP and EU F-Gas compliant refrigerants
  • Significant energy savings without compromising sample protection
  • Available in four upright sizes
  • Quiet operation
  • Ergonomically designed handle and eye-level display
  • Touchscreen interface puts information at your fingertips
Use our ULT selection guide, browse through our portfolio below, or ask one of our experts to help tailor a solution specific to your needs.
Quick links
Ultra-low upright freezers for exceptional performance every day
  300 Boxes* 400 Boxes 500 Boxes 600 Boxes
  300 box Forma ULT freezer 400 box Forma ULT freezer 500 box Forma ULT freezer 600 box Forma ULT freezer
Request a Quote
Capacity 14.9 cu. ft. (422L) 19.4 cu. ft. (549L) 24.1 cu. ft. (682L) 28.8 cu. ft. (816L)
Inner Doors 4 4 4 4
2mL Vial Capacity 30,000 40,000 50,000 60,000
Interior (DxWxH) 28.3 x 17.8 x 51.2 in.
(71.9 x 45.2 x 130.1cm)
28.3 x 23.1 x 51.2 in.
(71.9 x 58.8 x 130.1cm)
28.3 x 28.8 x 51.2 in.
(71.9 x 73.0 x 130.1cm)
28.3 x 34.4 x 51.2 in.
(71.9 x 87.3 x 130.1cm)
Exterior (DxWxH) 37.6 x 23 x 78 in.
(95.5 x 58.4 x 198.1cm)
37.6 x 28.3 x 78 in.
(95.5 x 71.9 x 198.1cm)
37.6 x 34.0 x 78 in.
(95.5 x 86.4 x 198.1cm)
37.6 x 39.6 x 78 in.
(95.5 x 100.6 x 198.1cm)
Shipping Weight 712 lbs. (323kg) 687 lbs. (312kg) 734 lbs. (333kg) 820 lbs. (372kg)
  300 Boxes* 400 Boxes 500 Boxes 600 Boxes
  300 box Revco ULT freezer 400 box Revco ULT freezer 500 box Revco ULT freezer 600 box Revco ULT freezer
Request a Quote
Capacity 14.9 cu. ft. (422L) 19.4 cu. ft. (549L) 24.1 cu. ft. (682L) 28.8 cu. ft. (816L)
Inner Doors 5 5 5 5
2mL Vial Capacity 30,000 40,000 50,000 60,000
Interior (DxWxH) 28.3 x 17.8 x 51.2 in.
(71.9 x 45.2 x 130.1cm)
28.3 x 23.1 x 51.2 in.
(71.9 x 58.8 x 130.1cm)
28.3 x 28.8 x 51.2 in.
(71.9 x 73.0 x 130.1cm)
28.3 x 34.4 x 51.2 in.
(71.9 x 87.3 x 130.1cm)
Exterior (DxWxH) 37.6 x 23 x 78 in.
(95.5 x 58.4 x 198.1cm)
37.6 x 28.3 x 78 in.
(95.5 x 71.9 x 198.1cm)
37.6 x 34.0 x 78 in.
(95.5 x 86.4 x 198.1cm)
37.6 x 39.6 x 78 in.
(95.5 x 100.6 x 198.1cm)
Shipping Weight 712 lbs. (323kg) 687 lbs. (312kg) 734 lbs. (333kg) 820 lbs. (372kg)
  300 Boxes* 400 Boxes 500 Boxes 600 Boxes
  300 box TS ULT freezer 400 box TS ULT freezer 500 box TS ULT freezer 600 box TS ULT freezer
Request a Quote
Capacity 14.9 cu. ft. (422L) 19.4 cu. ft. (549L) 24.1 cu. ft. (682L) 28.8 cu. ft. (816L)
Inner Doors 4 4 4 4
2mL Vial Capacity 30,000 40,000 50,000 60,000
Interior (DxWxH) 28.3 x 17.8 x 51.2 in.
(71.9 x 45.2 x 130.1cm)
28.3 x 23.1 x 51.2 in.
(71.9 x 58.8 x 130.1cm)
28.3 x 28.8 x 51.2 in.
(71.9 x 73.0 x 130.1cm)
28.3 x 34.4 x 51.2 in.
(71.9 x 87.3 x 130.1cm)
Exterior (DxWxH) 37.6 x 23 x 78 in.
(95.5 x 58.4 x 198.1cm)
37.6 x 28.3 x 78 in.
(95.5 x 71.9 x 198.1cm)
37.6 x 34.0 x 78 in.
(95.5 x 86.4 x 198.1cm)
37.6 x 39.6 x 78 in.
(95.5 x 100.6 x 198.1cm)
Shipping Weight 712 lbs. (323kg) 687 lbs. (312kg) 734 lbs. (333kg) 820 lbs. (372kg)
  300 Boxes* 400 Boxes 500 Boxes 600 Boxes
  300 box HERAfreeze ULT freezer 400 box HERAfreeze ULT freezer 500 box HERAfreeze ULT freezer 600 box HERAfreeze ULT freezer
Request a Quote
Capacity 14.9 cu. ft. (422L) 19.4 cu. ft. (549L) 24.1 cu. ft. (682L) 28.8 cu. ft. (816L)
Inner Doors
5 5 5 5
2mL Vial Capacity 30,000 40,000 50,000 60,000
Interior (DxWxH) 28.3 x 17.8 x 51.2 in.
(71.9 x 45.2 x 130.1cm)
28.3 x 23.1 x 51.2 in.
(71.9 x 58.8 x 130.1cm)
28.3 x 28.8 x 51.2 in.
(71.9 x 73.0 x 130.1cm)
28.3 x 34.4 x 51.2 in.
(71.9 x 87.3 x 130.1cm)
Exterior (DxWxH) 37.6 x 23 x 78 in.
(95.5 x 58.4 x 198.1cm)
37.6 x 28.3 x 78 in.
(95.5 x 71.9 x 198.1cm)
37.6 x 34.0 x 78 in.
(95.5 x 86.4 x 198.1cm)
37.6 x 39.6 x 78 in.
(95.5 x 100.6 x 198.1cm)
Shipping Weight 712 lbs. (323kg) 687 lbs. (312kg) 734 lbs. (333kg) 820 lbs. (372kg)
  300 Boxes* 400 Boxes 500 Boxes 600 Boxes
  300 box Forma ULT freezer 400 box Forma ULT freezer 500 box Forma ULT freezer 600 box Forma ULT freezer
Request a Quote
Capacity 14.9 cu. ft. (422L) 19.4 cu. ft. (549L) 24.1 cu. ft. (682L) 28.8 cu. ft. (816L)
Inner Doors 4 4 4 4
2mL Vial Capacity 30,000 40,000 50,000 60,000
Interior (DxWxH) 28.3 x 17.8 x 51.2 in.
(71.9 x 45.2 x 130.1cm)
28.3 x 23.1 x 51.2 in.
(71.9 x 58.8 x 130.1cm)
28.3 x 28.8 x 51.2 in.
(71.9 x 73.0 x 130.1cm)
28.3 x 34.4 x 51.2 in.
(71.9 x 87.3 x 130.1cm)
Exterior (DxWxH) 37.6 x 23 x 78 in.
(95.5 x 58.4 x 198.1cm)
37.6 x 28.3 x 78 in.
(95.5 x 71.9 x 198.1cm)
37.6 x 34.0 x 78 in.
(95.5 x 86.4 x 198.1cm)
37.6 x 39.6 x 78 in.
(95.5 x 100.6 x 198.1cm)
Shipping Weight 712 lbs. (323kg) 687 lbs. (312kg) 734 lbs. (333kg) 820 lbs. (372kg)
  300 Boxes* 400 Boxes 500 Boxes 600 Boxes
  300 box Revco ULT freezer 400 box Revco ULT freezer 500 box Revco ULT freezer 600 box Revco ULT freezer
Request a Quote
Capacity 14.9 cu. ft. (422L) 19.4 cu. ft. (549L) 24.1 cu. ft. (682L) 28.8 cu. ft. (816L)
Inner Doors 5 5 5 5
2mL Vial Capacity 30,000 40,000 50,000 60,000
Interior (DxWxH) 28.3 x 17.8 x 51.2 in.
(71.9 x 45.2 x 130.1cm)
28.3 x 23.1 x 51.2 in.
(71.9 x 58.8 x 130.1cm)
28.3 x 28.8 x 51.2 in.
(71.9 x 73.0 x 130.1cm)
28.3 x 34.4 x 51.2 in.
(71.9 x 87.3 x 130.1cm)
Exterior (DxWxH) 37.6 x 23 x 78 in.
(95.5 x 58.4 x 198.1cm)
37.6 x 28.3 x 78 in.
(95.5 x 71.9 x 198.1cm)
37.6 x 34.0 x 78 in.
(95.5 x 86.4 x 198.1cm)
37.6 x 39.6 x 78 in.
(95.5 x 100.6 x 198.1cm)
Shipping Weight 712 lbs. (323kg) 687 lbs. (312kg) 734 lbs. (333kg) 820 lbs. (372kg)
  300 Boxes* 400 Boxes 500 Boxes 600 Boxes
  300 box TS ULT freezer 400 box TS ULT freezer 500 box TS ULT freezer 600 box TS ULT freezer
Request a Quote
Capacity 14.9 cu. ft. (422L) 19.4 cu. ft. (549L) 24.1 cu. ft. (682L) 28.8 cu. ft. (816L)
Inner Doors 4 4 4 4
2mL Vial Capacity 30,000 40,000 50,000 60,000
Interior (DxWxH) 28.3 x 17.8 x 51.2 in.
(71.9 x 45.2 x 130.1cm)
28.3 x 23.1 x 51.2 in.
(71.9 x 58.8 x 130.1cm)
28.3 x 28.8 x 51.2 in.
(71.9 x 73.0 x 130.1cm)
28.3 x 34.4 x 51.2 in.
(71.9 x 87.3 x 130.1cm)
Exterior (DxWxH) 37.6 x 23 x 78 in.
(95.5 x 58.4 x 198.1cm)
37.6 x 28.3 x 78 in.
(95.5 x 71.9 x 198.1cm)
37.6 x 34.0 x 78 in.
(95.5 x 86.4 x 198.1cm)
37.6 x 39.6 x 78 in.
(95.5 x 100.6 x 198.1cm)
Shipping Weight 712 lbs. (323kg) 687 lbs. (312kg) 734 lbs. (333kg) 820 lbs. (372kg)
  300 Boxes* 400 Boxes 500 Boxes 600 Boxes
  300 box HERAfreeze ULT freezer 400 box HERAfreeze ULT freezer 500 box HERAfreeze ULT freezer 600 box HERAfreeze ULT freezer
Request a Quote
Capacity 14.9 cu. ft. (422L) 19.4 cu. ft. (549L) 24.1 cu. ft. (682L) 28.8 cu. ft. (816L)
Inner Doors
5 5 5 5
2mL Vial Capacity 30,000 40,000 50,000 60,000
Interior (DxWxH) 28.3 x 17.8 x 51.2 in.
(71.9 x 45.2 x 130.1cm)
28.3 x 23.1 x 51.2 in.
(71.9 x 58.8 x 130.1cm)
28.3 x 28.8 x 51.2 in.
(71.9 x 73.0 x 130.1cm)
28.3 x 34.4 x 51.2 in.
(71.9 x 87.3 x 130.1cm)
Exterior (DxWxH) 37.6 x 23 x 78 in.
(95.5 x 58.4 x 198.1cm)
37.6 x 28.3 x 78 in.
(95.5 x 71.9 x 198.1cm)
37.6 x 34.0 x 78 in.
(95.5 x 86.4 x 198.1cm)
37.6 x 39.6 x 78 in.
(95.5 x 100.6 x 198.1cm)
Shipping Weight 712 lbs. (323kg) 687 lbs. (312kg) 734 lbs. (333kg) 820 lbs. (372kg)
*Box capacity is in terms of 2-inch (5cm) high boxes.
Top
Configure your ultra-low storage solution
Find an ultra-low temperature freezer by walking through a series of selections based on your lab space requirements. A representative will contact you soon with a recommendation.
Find your freezer
Which plug type do I need?
  Standard Plug Type Voltage Hz
NEMA 5-15 115–120 60
NEMA 5-20 115–120 60
NEMA 6-15 208–230 60
Continental EU
230 50
Alternative plugs and cords are available to meet your special needs. Learn more ›
Top
Monitor your sample storage conditions 24/7
Smart-vue-monitor-mobile-application-278x195
In the event of a power or mechanical issue, you need to be notified right away. Our wireless monitoring systems are designed for 24/7 monitoring of your ultra-low freezer's performance along with reporting, remote alarm notification and continuous data collection.
View our wireless monitoring solutions ›
Cryogenic storage
Matrix storage (barcoded tubes)
Cryo extra sample prep
Matrix Tubes
Standard storage tubes
Racks, boxes and accessories
Externally threaded universal tubes
Freezer racks and boxes
Top
How-to freezer videos
Top