46610-Fluor-Biotin-Quant-230
The simple and accurate HABA dye assay and fluorescent detection kits are used to measure the amount of biotin attached to proteins or other biomolecules following biotinylation. Now you can determine the molar ratio of biotin to labeled antibody molecule in an easy, efficient manner.
Choose the biotin quantitation kits or reagents for your experiment
Kit categories Product Product names Product features Detection method
Fluorescence Biotin Quantitation Kit 46610 Fluorescence Biotin Quantitation Kit Fluorescent assay reagent and HABA dye for rapid and sensitive determination of biotinylation levels in labeled antibodies and other proteins with minimal sample quantities Fluorescent
Pierce Biotin Quantitation Kit 28005 Pierce Biotin Quantitation Kit A simple, colorimetric method for determining biotinylation efficiency Colorimetric
HABA Reagent 28010 HABA HABA dye reagent and biotinylated protein standard for convenient colorimetric determination of biotinylation levels in antibodies and other proteins Colorimetric