Barcode Readers
A full storage and handling product line
From 2D barcode readers to sample handling software, from storage tubes to cappers/decappers and sealers, we offer a full storage and handling product line to enhance your throughput of manual or integrated workflows.
Performance features of Barcode Readers
Features | VisionMate ST Single Tube 2D Reader | VisionMate Wireless Reader |
VisionMate SR Single Rack 2D Reader | VisionMate High Speed Barcode Reader |
![]() |
![]() |
![]() |
![]() |
|
Scan 2D Tube Scan | X | X | X | |
Scan 96-way rack of 2D tubes | X | X | ||
Scan 48-way rack of 2D tubes | X | X | ||
Scan 24-way rack of 2D tubes | X | X | ||
Read 2D orientation plug on bottom of rack | X | X | ||
Automatic Rack Detection and Scan | X | |||
Automatic Rack Orientation | X | |||
Rack Linear Barcode Scan | X | X | ||
Automatic Linear Barcode Detection | X | |||
Fast scanning (less than 5 seconds) |
X | X | X | |
CSV, XLS data export | X | X | X | X |
Keyboard Wedge Option | X | X | X | X |
TCIP data export (preffered for LIMS) |
X | |||
Integration friendly | X | X | ||
Order Now | Order Now | Order Now | Order Now |
Compatibility with Matrix Tubes
Our line of Barcode Readers is compatible with Matrix tubes. The linear and human readable codes match this unique 2D barcode, allowing 1D scanning or visual identification of samples across satellite or collaborative sites without 2D readers.
Select a video to learn more about the Wireless Barcode Reader
Setting Up and Installing the Thermo Scientific VisionMate Wireless Barcode Reader
How to Use the VisionMate Wireless Barcode Reader Device and Software for Data Export
Don't forget about the software.
Learn more about our VisionMate software for our sample storage product line.
Resources
Support
Unity Lab Services: Access to our expert service team, information on support plans, parts and consumables resources, and other instrument services.