Sanger sequencing and fragment analysis end-to-end workflows, high performance solutions
Created by a team of global professionals and built upon decades of proven results, Applied Biosystems genetic analyzers, consumables and software help optimize your Sanger sequencing and fragment analysis workflows with precision. These fast and simple workflows enable the completion of your targeted sequencing interrogation, from sample to answer, in less than a workday.
Fragment analysis workflow
Explore the process of separating fluorescently labeled DNA fragments through capillary electrophoresis (CE) and determining their sizes by comparing them to a size standard.
Sanger sequencing workflow
On-instrument consumables
The SeqStudio cartridge has an integrated system design that includes a capillary array, Pop-1 universal polymer, buffer, and pump.
Check out our wide selection of CE consumables ›
Video: How to perform Sanger sequencing
Fragment analysis workflow
On-instrument consumables
The SeqStudio cartridge has an integrated system design that includes a capillary array, Pop-1 universal polymer, buffer, and pump.
Check out our wide selection of CE consumables of CE consumables ›
Primer Designer tool
Use our online Primer Designer Tool to search for the right PCR/Sanger sequencing primer pair from a database of ~650,000 predesigned primer pairs for resequencing the human exome and human mitochondrial genome.
Check out our Primer designer tool ›
Video: How does Fragment analysis work?
Whether your laboratory demands top-tier service levels and strict compliance with regulatory guidelines or you aim to optimize performance within a constrained budget or anything in between, we provide a variety of service plans tailored to meet your specific requirements.
Style Sheet for Global Design System
innovigene_900x505
InnoviGene Suite
InnoviGene Suite is an all-in-one, browser-based platform that offers a suite of Sanger sequencing applications software, providing a harmonious bouquet of tools to automate and optimize your data analysis.
Explore InnoviGene Suite ›
dynabeads_900x505
Dynabeads Sequencing Cleanup Kit
The Dynabeads Sequencing Cleanup Kit uses uniform, superparamagnetic beads to rapidly and effectively purify dye-terminator sequencing reactions, enhancing the efficiency and reliability of Sanger sequencing-based protocols.
Learn more ›