Manganese • Transition Metal
Primary XPS region: Mn2p
Overlapping regions: Cu LMM, Au4p1/2
Binding energies of common chemical states:
Chemical stateBinding energy Mn2p3/2
Mn metal638.7 eV
MnO641.4 eV
Mn2O3641.4 eV
MnO2641.8 eV
Experimental information
  • Collect Mn3s region to enable straightforward identification of Mn oxidation state.
  • Some compounds/alloys of manganese are magnetic.
    • Degauss samples if possible before analysis.
Interpretation of XPS spectra
  • Mn2p peak has significantly split spin-orbit components (Δmetal=11.2eV).
  • Mn2p peaks have asymmetric shape for metal.
    • Mn oxide peaks are significantly shifted to higher binding energy than the metal.
    • A well resolved Mn2p spectrum for metal shows multiplet splitting [1].
      • Second component (shifted by 0.9eV from main peak) can be neglected when fitting mixed oxide/metal spectra.
High Resolution Mn2p3/2 Spectrum Manganese Metal
  • Method for chemical analysis of Mn oxides:
    • Mn2p peaks for Mn oxides have many multiplet-split components, similar to the Cr2p region in chromium oxides.
    • MnO2 has a much narrower Mn2p3/2 peak compared to MnO or Mn2O3.
      • Also has an identifiable shape at the top of the Mn2p3/2 peak.
    • MnO has a satellite feature (~647eV), which is not present for either Mn2O3 or MnO2.
Mn2p Spectra Mn Oxides
  • It is possible to distinguish Mn oxidation states using Mn3s peak.
    • This peak has two multiplet split components.
      • Caused by coupling of non-ionized 3s electron with 3d valence-band electrons.
    • Magnitude of peak splitting is diagnostic of oxidation state.
      • ΔE for MnO (Mn2+) = 6.0eV, Mn2O3 (Mn3+) ≥ 5.3eV, MnO2 (Mn4+) = 4.7eV.
Mn3s Spectra Mn Oxides
References
  • [1] MC Biesinger et al., Applied Surface Science 257 (2011) 2717-2730.
Get materials science and electron microscopy updates
Manganese • Transition Metal
Element Crystal BCC
About the element
Symbol: Mn
Date of discovery: 1774
Name origin: Latin magnes
Appearance: silverish/grayish
Discoverer: Johann Gahn
Obtained from: brine, sea water
Melting point: 1519 K
Boiling point: 2334 K
Density[kg/m3]: 7470
Molar volume: 7.35 × 10-6 m3/mol
Protons/Electrons: 25
Neutrons: 30
Shell structure: 2,8,13,2
Electron configuration: [Ar]3d54s2
Oxidation state: 7,6,4,2,3
Crystal structure: cubic body centered
Manganese is used in the same disciplines today as it was in prehistoric times. Used in iron and steel production, it was found in iron ores forged by the Spartans. The Egyptians and Romans used manganese compounds to add or remove color from glass. Manganese compounds can color glass an amethyst color and produce the color of the gemstone amethyst. Most of the world’s deposits of manganese are found in South Africa and Ukraine. Manganese also exists in nodules on the ocean floor; however, no economical method of harvesting them has been found. Potassium permanganate (KMnO4) is commonly used in laboratories for its oxidizing properties.
Products
Nexsa XPS
  • Módulo de inclinação para medições ARXPS
  • Fonte de íons de modo duplo para recursos de perfil de profundidade expandida
  • Análise de isolantes
Folha de estilo para cartões de instrumentos original
K-Alpha XPS
  • Espectroscopia de área selecionável
  • Monocromador microfocalizado
  • Espectroscopia de estado químico de alta resolução
ESCALAB Xi+ XPS
  • Espectroscopia de alta sensibilidade
  • XPS com raios X não monocromáticos
  • Analisador de energia hemisférica de 180 graus
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Folha de estilo para especificações de tabela de produtos
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Folha de estilo para especificações de tabela de produtos
Fix for left nav
Folha de estilo para Rodapé de suporte e serviço
Folha de estilo para Fontes
Folha de estilo para Cartões
Serviços de microscopia eletrônica para
a ciência dos materiais
Para garantir o desempenho ideal do sistema, fornecemos acesso a uma rede de especialistas em serviços de campo, suporte técnico e peças de reposição certificadas.