Ruthenium • Transition Metal
Primary XPS region: Ru3d
Overlapping regions: C1s
Binding energies of common chemical states:
Chemical stateBinding energy Ru3d5/2
Ru metal280.2 eV
RuO2280.7 eV
Oxide charge referenced to adventitious C1s peak at 284.8eV.
Experimental information
N/A
Interpretation of XPS spectra
  • Ru3d region has significantly split spin-orbit components (D=4.2eV).
    • Ru metal and RuO2 both give asymmetric Ru3d peaks shapes.
    • For ruthenium metal and RuO2, the Ru3d3/2 component is shorter than expected (for a d5/2, d3/2 doublet) and is much broader than the Ru3d5/2 component.
  • There is a very strong overlap between the Ru3d and C1s regions.
    • If possible, peak fit a clean standard ruthenium material to develop the fitting methodology and then add C1s peaks as required (see below).
element-ruthenium-600x400
About this element
Element Crystal hex
Symbol: Ru
Date of discovery: 1844
Name origin: Latin Ruthenia
Appearance: silvery
Discoverer: Karl Karlovich Klaus
Obtained from: pentlandite, pyroxinite
Melting point: 2607 K
Boiling point: 4423 K
Density[kg/m3]: 12370
Molar volume: 8.17 × 10-6 m3/mol
Protons/Electrons: 44
Neutrons: 57
Shell structure: 2,8,18,15,1
Electron configuration: [Kr]4d75s1
Oxidation state: 2,3,4,6,8
Crystal structure: hexagonal
Ruthenium is a member of the platinum group, which also includes rhodium, palladium, osmium, iridium, and (of course) platinum. Ruthenium rapidly oxidizes, but does not tarnish in normal temperatures. Small amounts of ruthenium can enhance the properties of other elements. Platinum and palladium become harder, while titanium is more resistant to corrosion after the addition of ruthenium. Ruthenium also catalyzes the splitting of hydrogen sulfide and other chemical processes. Ruthenium may have anti-cancer medical applications, as well. This element is primarily found in the Ural Mountains and North and South America.
elements_ruthenium
Products
Nexsa G2 XPS
  • Micro-focus X-ray sources
  • Unique multi-technique options
  • Dual-mode ion source for monoatomic & cluster ion depth profiling
Style Sheet for Instrument Cards Original
K-Alpha XPS
  • High resolution XPS
  • Fast, efficient, automated workflow
  • Ion source for depth profiling
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Electron microscopy services for
the materials science
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.