CoverWell Chamber Gaskets—Table 23.6
Table 23.6 CoverWell chamber gaskets.
| Cat # | Number of Wells | Well Dimensions | Depth | Approximate Volume Per Chamber | Quantity per Package |
|---|---|---|---|---|---|
| CoverWell incubation chamber gaskets | |||||
| C18150 | 1 | 40 mm × 22 mm | 0.2 mm | 200 µL | 25 |
| C18151 | 1 | 40 mm × 22 mm | 0.5 mm | 500 µL | 50 |
| C18155 | 1 | 13 mm diameter | 0.2 mm | 20 µL | 25 |
| C18156 | 1 | 13 mm diameter | 0.5 mm | 20 µL | 50 |
| CoverWell imaging chamber gaskets | |||||
| C18160 | 1 | 20 mm diameter | 0.5 mm | 180 µL | 40 |
| C18161 | 1 | 20 mm diameter | 1.0 mm | 300 µL | 40 |
| C24726 * | 1 | 20 mm diameter | 0.5 mm | 180 µL | 40 |
| C24727 * | 1 | 20 mm diameter | 1.0 mm | 300 µL | 40 |
| CoverWell perfusion chamber gaskets | |||||
| C18120 | 1 | 32 mm × 19 mm | 0.5 mm | 350 µL | 40 |
| C18121 | 1 | 32 mm × 19 mm | 1.0 mm | 550 µL | 40 |
| C18128 | 4 | 19 mm × 6 mm | 0.5 mm | 70 µL | 40 |
| C18136 | 1 | 20 mm diameter | 1.0 mm | 300 µL | 40 |
| C18139 | 8 | 9 mm diameter | 0.5 mm | 35 µL | 20 |
| C18140 | 8 | 9 mm diameter | 1.0 mm | 60 µL | 20 |
| C18141 | 8 | 9 mm diameter | 2.0 mm | 100 µL | 20 |
| C18142 | 8 | 9 mm diameter | 2.5 mm | 150 µL | 20 |
| * With adhesive on one side. | |||||
For Research Use Only. Not for use in diagnostic procedures.