Search
Western blot analysis was performed on tissue extracts (30 ug lysate) of Mouse Muscle (Lane 1), and Mouse Heart (lane 2). The blots were probed with Anti-Junctophilin-1 Rabbit Polyclonal Antibody(Product# 405100, 2 ug/mL) and detected by chemiluminescence Goat Anti-Rabbit IgG Secondary Antibody, HRP conjugate(Product # G21234,1:5000 dilution). A 70 kDa band corresponding to Junctophilin-1 was observed across cell lines tested. Known quantity of protein samples were electrophoresed using Novex® NuPAGE® 10 % Bis-Tris gel (Product # NP0301BOX), XCell SureLock™ Electrophoresis System (Product # EI0002) and Novex® Sharp Pre-Stained Protein Standard (Product # LC5800). Resolved proteins were then transferred onto a nitrocellulose membrane by iBlot® 2 Dry Blotting System (Product # IB21001).The membrane was probed with the relevant primary and secondary Antibody following blocking with 5 % skimmed milk. Chemiluminescent detection was performed using Novex® ECL Chemiluminescent Substrate Reagent Kit (Product # WP20005).