Chromatin immunoprecipitations were performed with cross-linked chromatin from 4 x 106 HeLa cells and either 20 ?L of Acetyl-Histone H4 (Lys5) Polyclonal Antibody or 2 ?L of normal rabbit IgG. The enriched DNA was quantified by real-time PCR. The amount of immunoprecipitated DNA in each sample is represented as signal relative to the total amount of input chromatin, which is equivalent to one.
Related Products
Related Images