On this page:
High-throughput analysis of recombinant proteins and antibodies with POROS HPLC Columns
Thermo Scientific POROS HPLC columns offer a highly efficient solution for the analysis of recombinant proteins and antibodies. These general purpose columns have various modes of separation—anion exchange, cation exchange, and reversed phase—and are designed for high-throughput analysis along the bioprocessing workflow. Ionic polymer ligands bound to a robust POROS bead facilitate exceptional performance in high-throughput and high-specificity applications. Ideal for use in biotechnology and pharmaceutical research and manufacturing, these columns offer several key benefits:
  • High binding capacity: Designed for robust analysis over a wide sample load of recombinant proteins
  • High-specificity analysis: Offers precise and detailed characterization of recombinant proteins
  • Rapid mass transfer: Enables faster analysis times and higher throughput
  • Exceptional mechanical stability: Supports high flow rates and robust performance in demanding applications
  • Versatility: Suitable for a wide range of recombinant proteins and can be utilized in various research and development processes
  • Reusability: Designed for multiple cycles for cost-effective and reproducible quantitation over hundreds of chromatography cycles
POROS bead technology
Thermo Scientific POROS bead technology revolutionizes biomolecule purification and analysis with its robust polymer matrix, offering exceptional mechanical stability and high flow rates for HPLC applications. The unique structure of POROS beads creates a high surface area, facilitating exceptional binding capacity and rapid mass transfer, ultimately contributing to faster processing times and higher throughput. Its versatility supports a wide range of ligands, making it an indispensable and powerful technology for applications in biotechnology, pharmaceuticals, and life sciences.
Ion exchange chromatography
Ion exchange chromatography is a method used to separate ionic molecules such as proteins based on their charge. In this technique, a sample containing the molecule of interest is injected into the chromatography system and passed through a column filled with a resin that has charged functional groups. These groups interact with the charged molecules in the sample. A liquid mobile phase is used to carry the sample through the column, and the ions are separated based on their affinity to the resin. A counter ion such as sodium or chloride is used to displace or exchange the molecule bound to the surface of the resin. The concentration of counter ion needed depends on how strongly the molecule binds to the resin. As the molecules elute from the column, they are detected and quantified.
POROS 10 and 20 µm strong anion exchange columns
POROS HQ Columns
POROS 20 µm weak anion exchange columns
POROS PI
POROS 20 µm cation exchange columns
Reversed-phase chromatography
Reversed phase chromatography is used to separate non-polar compounds. In this method, the sample is injected into the chromatography system and passed through a column filled with a non-polar stationary phase, typically hydrophobic polymer or silica particles coated with hydrophobic groups. A polar mobile phase, usually a mixture of water and an organic solvent like methanol or acetonitrile, is used to carry the sample through the column. Non-polar compounds interact more strongly with the stationary phase and are retained longer, allowing for their separation. The compounds are then detected and quantified as they elute from the column.
POROS 10 and 20 µm reversed-phase columns
Oligo R3 is a 30 µm Poly[styrenedivinylbenzene] particle designed for the reversed-phase separation of polynucleotides
Request a technical consultation
No matter your questions, we are here to help. Our experienced team of scientists and technical specialists is ready to assist you in choosing the right solution and products. Complete this form to start the conversation.
Analytics Knowledge Hub
Explore the Analytics Knowledge Hub and discover a robust collection of webinars, articles, eBooks, infographics, and other digital resources to help you optimize analytical testing in your bioprocess workflow.