Quickly obtain composite materials' properties from your images
Composite materials are making their way into many different application areas, ranging from aerospace to automotive and construction. Their properties improve stiffness and strength and allow for design of lightweight components at a reasonable cost. A composite is commonly defined as a combination of two or more distinct materials, each of which retains its own distinctive properties, to create a new material with properties that cannot be achieved by any of the components acting alone.
Thermo Scientific Avizo Software allows you to obtain structural, physical, chemical, and mechanical properties quickly and accurately. With dedicated tools to obtain fiber properties, porosity, and pore network information, Avizo Software is the digital analytic lab you need to analyze your 2D and 3D imaging data, no matter the data acquisition system you used.
Imaging data
Workflow using Avizo Software
Processing
Workflow using Avizo Software
Visualization
Workflow using Avizo Software
Analysis
Workflow using Avizo Software
Customization
Confidence
Thanks to powerful segmentation and image processing capabilities and workflows, and after 20+ years of collaboration with the scientific community and thousands of researchers, it has been now proven that our digital imaging-based workflows provide reliable answers to industrial and scientific problems.
Customization
Customization
Because your needs are unique and keep evolving, our software solutions are fully flexible and open to customization. Thanks to our scripting interface (Python, TCL), bridge with MATLAB, and our programming API, you can expand our software solution and integrate your own IP (Intellectual Property). And, if needed, our professional service team can help you design unique solutions tailored to your needs.
icon-improve_collaboration_64x64
Support
Thanks to our dedicated professional support team, you get access to our top experts to ensure that no question is left unanswered. And with our training, consulting options, and ever-expanding collection of tutorials and how-to’s, you can reduce your learning curve and focus on getting the answers you are pursuing.
Software_Automation_65x64
Automation
Thanks to our automation capabilities, and with the addition of our always expanding online repository of add-ons (Xtra Gallery), you can encapsulate repeatable workflows into easy-to-reproduce recipes. With the addition of artificial intelligence, analysis can be performed by non-image processing experts, allowing them to save time on complex analysis while ensuring results consistency. 
Particle-reinforced use cases library
Particle reinforced
composites
Learn more ›
Fiber-reinforced use cases library
Fiber reinforced
composites
Learn more ›
Dispersion-strengthened composites use cases library
Dispersion strengthened composites
Learn more ›
On-demand webinar: Imaging data management with Athena Software
On-demand webinar: “Introduction of Avizo Software for Composites Characterization”
Learn how Thermo Scientific Avizo Software can enable you to:
  • Accelerate design and development cycles for optimized manufacturing of composite materials
  • Apply a digital workflow for characterization and failure analysis of composites
  • Anticipate causes of failures through better assertion of manufactured parts
  • Speed up the design of coating materials and reduce failure rates
Watch now
Techniques
Fiber analysis
Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of fibrous materials. Avizo Software enables the detection of manufacturing process damage, such as matrix cracking, fiber/matrix debonding, or fiber breakage. Defects in the material can be quantified locally (voids, cracks) or globally (cross-sectional distortions or misalignment).
Learn more
Porosity analysis
Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of porous materials. Avizo Software enables the detection and classification of various types of porosity (for example, connected vs isolated; macro pores vs micro pores), even on images with complex artifacts (such as pore back effect in FIB-SEM).
Learn more
Pore Network Analysis
Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of porous materials. This extension enables the conversion of the pore space into a model (Pore Network Model) representing pores and their connectivity, on which various properties can be color mapped. It also provides advanced pore space statistics (pore size distribution, pore throat size distribution, channel length, etc.) and filtering of the model based on its properties.
Permeability analysis
Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of your materials. The Xlab extension provides numerical simulation capabilities to compute absolute permeability, thermal conductivity, electrical resistivity, and molecular diffusivity.
Failure Analysis
Avizo Software provides a fast and robust meshing engine to automatically create simulation-ready meshes from 3D labeled images. Avizo Software generates high-quality meshes with smoothed conforming interfaces from large multi-material segmented images. You have fine control over element size distribution, while Avizo Software preserves thin structures and topology. Meshes can be exported to major solver’s format along with boundary conditions. Avizo Software also provides import of the solver’s outputs for post-processing the results with advanced visualization tools.
Grain Analysis
Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of grains. Avizo Software provides advanced tools and enables the detection, segmentation, and analysis of grains, their size, distribution, and shape, even on complex images with artifacts. Grains can then be filtered, classified, and analyzed allowing full size, distribution, and shape statistical extraction. Avizo Software also provides the tools to automate and repeat your analysis through easy-to-customize recipes.
Particles analysis
Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of particles to improve performance of your materials, whether catalysts, alloys, or polymers. Avizo Software provides advanced tools to enable the detection, segmentation, and analysis of particles. Particles can then be filtered, classified, and analyzed, allowing full size, distribution, and shape statistical extraction. Avizo Software also provides the tools to automate and repeat your analysis through easy-to-customize recipes.
Fiber analysis
Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of fibrous materials. Avizo Software enables the detection of manufacturing process damage, such as matrix cracking, fiber/matrix debonding, or fiber breakage. Defects in the material can be quantified locally (voids, cracks) or globally (cross-sectional distortions or misalignment).
Learn more
Porosity analysis
Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of porous materials. Avizo Software enables the detection and classification of various types of porosity (for example, connected vs isolated; macro pores vs micro pores), even on images with complex artifacts (such as pore back effect in FIB-SEM).
Learn more
Pore Network Analysis
Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of porous materials. This extension enables the conversion of the pore space into a model (Pore Network Model) representing pores and their connectivity, on which various properties can be color mapped. It also provides advanced pore space statistics (pore size distribution, pore throat size distribution, channel length, etc.) and filtering of the model based on its properties.
Permeability analysis
Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of your materials. The Xlab extension provides numerical simulation capabilities to compute absolute permeability, thermal conductivity, electrical resistivity, and molecular diffusivity.
Failure Analysis
Avizo Software provides a fast and robust meshing engine to automatically create simulation-ready meshes from 3D labeled images. Avizo Software generates high-quality meshes with smoothed conforming interfaces from large multi-material segmented images. You have fine control over element size distribution, while Avizo Software preserves thin structures and topology. Meshes can be exported to major solver’s format along with boundary conditions. Avizo Software also provides import of the solver’s outputs for post-processing the results with advanced visualization tools.
Grain Analysis
Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of grains. Avizo Software provides advanced tools and enables the detection, segmentation, and analysis of grains, their size, distribution, and shape, even on complex images with artifacts. Grains can then be filtered, classified, and analyzed allowing full size, distribution, and shape statistical extraction. Avizo Software also provides the tools to automate and repeat your analysis through easy-to-customize recipes.
Particles analysis
Avizo Software is an all-in-one image analysis platform for the visualization, processing, and quantification of particles to improve performance of your materials, whether catalysts, alloys, or polymers. Avizo Software provides advanced tools to enable the detection, segmentation, and analysis of particles. Particles can then be filtered, classified, and analyzed, allowing full size, distribution, and shape statistical extraction. Avizo Software also provides the tools to automate and repeat your analysis through easy-to-customize recipes.
Services
Amira-Avizo-Introductory-training_1160x600
Introductory training
Shorten your learning curve and maximize your investment with this introductory training specifically designed for new users of Amira, Avizo and PerGeos Software.
The course consists of a lecture with hands-on sessions. The training material highlights the basic features and functionalities of Amira, Avizo and PerGeos Software.
Amira-Avizo-advanced-training_1160x600
Advanced training
Maximize your investment and reduce your time-to-results with this advanced training specifically designed for existing users of Amira, Avizo and PerGeos Software.
The course consists of a lecture with hands-on sessions. The training material highlights advanced features and functionalities of Amira, Avizo and PerGeos Software.
Amira-Avizo-Custom-Dev_1160x600
Custom development
With over 25 years of experience in 3D and image processing and hundreds of custom projects delivered to organizations small and large, Thermo Fisher Scientific can provide you with a solution tailored to fit your specific needs.
We can customize and expand our software solutions at various levels.
Register now to get exclusive content delivered directly to your inbox.
Register now
Resources
3D fiber reconstruction in fiber-reinforced concrete with Avizo Software
Fiber characterization and orientation analysis in CFRP with Avizo Software
Defect analysis of a CFRP bike frame with Avizo Software
Characterization and quantification in an additively manufactured scaffold with Avizo Software
Digital Volume Correlation on PA6 matrix reinforced with glass fibers using Avizo Software
Distribution of individual fiber orientations within a sample material
Experts from Thermo Fisher Scientific highlight three unique approaches that may operate at different resolutions to provide insights about the fiber orientations.
Read more ›
3D fiber reconstruction in fiber-reinforced concrete with Avizo Software
Fiber characterization and orientation analysis in CFRP with Avizo Software
Defect analysis of a CFRP bike frame with Avizo Software
Characterization and quantification in an additively manufactured scaffold with Avizo Software
Digital Volume Correlation on PA6 matrix reinforced with glass fibers using Avizo Software
Distribution of individual fiber orientations within a sample material
Experts from Thermo Fisher Scientific highlight three unique approaches that may operate at different resolutions to provide insights about the fiber orientations.
Read more ›
Features
Import and process your imaging data
    • Handle any modality, at any scale, of any size:
    - X-ray tomography: CT, micro-/nanoCT
    - Electron microscopy
    - Synchrotron
    • Support for multi-data/multi-view, multi-channel, time series, very large data
    • Scaling, calibration, conversion, re-sampling
    • Image enhancement, comprehensive filtering and convolution, Fourier frequency transforms
    • Artifact reduction algorithms
    • Advanced multi-mode 2D/3D automatic registration
    • Image stack alignment, arithmetic, correlation, fusion
    Easily segment your imaging data
    • Thresholding and auto-segmentation, object separation, automatic labeling
    • Region growing, snakes, interpolation, wrapping, smoothing
    • Morphological processing, including watershed and basins
    • Machine Learning-based segmentation
    • Automatic tracing of individual fibers and filaments
    • Skeletonization and filament network extraction
    • 3D surface reconstruction
    • Grid generation for FEA/CFD
    Image-to-simulation workflows
    • 3D image-based meshing for Finite Element and CFD
    • Porosity/pore connectivity analysis and skeletonization for Pore Network Modeling
    • Direct 3D image-based simulation: absolute permeability, molecular diffusivity, electrical resistivity, and thermal conductivity computation
    Export your analysis and visualization work to seamlessly publish and present it
    • Animation and video generation
    • Advanced key frame and object animation
    • Mix images, geometric models, measurements and simulations
    • Annotations, measures legends, histograms and curve plots
    • Export spreadsheets, 3D models, high-quality images
    Visualize and explore your imaging data
    • Interactive high-quality volume
    • Orthogonal, oblique, cylindrical and curved slicing
    • Contouring and iso-surface extraction
    • Maximum Intensity or other types of projections
    • Vector and tensor visualization
    Analyze your imaging data and obtain quantitative information
    • Intuitive recipe creation, customization, automated replay
    • Built-in measurements, including counts, volumes, areas, perimeters, aspect ratios and orientations
    • User-defined measures
    • Results viewer with spreadsheet tool and charting
    • Automatic individual feature measurements, 3D localization and spreadsheet selection
    • Automated statistics, distribution graphs
    • Feature filtering using any measurement criterion
    • Data registration, deformation, comparison and measurements
    • Porosity detection and measurement
    • Fiber analysis
    • Pre-processing for structural and flow simulations
    • Import of CAD models for actual/nominal comparison
    Easily and quickly adapt Avizo Software to your specific needs
    • Custom C++ modules development
    • MATLAB™ bridge
    • Python scripting API