Analytical instruments for automotive materials characterization
Automotive materials characterization typically relies on coordinated, multi-technique workflows to resolve complex structural and chemical questions. High-resolution imaging is often used first to locate defects, particles, or regions of interest. Elemental and particle analysis then provide quantitative information on composition, size distribution, and phase association. Surface-sensitive techniques help analyze the chemical state of coatings, interfaces, and contamination layers, while depth profiling reveals compositional changes across multilayer systems. Site-specific sample preparation supports subsequent high-resolution or nanoscale analysis, including three-dimensional reconstruction and crystallographic evaluation. By integrating results across these techniques, engineers can correlate microstructure, chemistry, and processing conditions, which supports robust root-cause failure analysis, process optimization, and standards-compliant quality control across automotive production workflows.
Scanning electron microscopes
Scanning electron microscopes offer high-resolution imaging, elemental analysis, and microstructural characterization of automotive materials and components. SEMs are widely used for particle and inclusion analysis, failure analysis, quality control, and process optimization. Integrated elemental mapping rapidly identifies contamination, phase distribution, and surface defects. Advanced SEM platforms also support automated and statistical workflows for technical cleanliness, making them well suited to production and quality environments.
Thermo Scientific ChemiSEM Systems combine high-resolution SEM imaging with real-time elemental analysis, delivering immediate chemical insight without interrupting imaging workflows. These systems are particularly effective for technical cleanliness, inclusion analysis, coating evaluation, corrosion studies, and failure analysis. Live elemental feedback supports faster decision-making and consistent results in both research and manufacturing environments.
The Thermo Scientific TruePix EBSD Detector provides high-speed, high-sensitivity crystallographic analysis for comprehensive microstructural characterization of automotive materials. The system offers accurate phase identification, grain structure measurement, texture analysis, and deformation studies across a wide range of metals and alloys. The TruePix EBSD Detector also delivers reliable indexing in complex and multiphase materials, supporting detailed insight into material performance and processing effects. Fully integrated with the Thermo Scientific Apreo ChemiSEM System, it supports automated, statistically robust workflows for research, quality control, and failure analysis in automotive development and manufacturing environments.
SEM techniques for automotive materials analysis
  • Elemental analysis
  • Particle and inclusion analysis
  • Surface analysis
  • Phase and microstructure analysis
  • Automated and statistical analysis
Automotive use cases of SEM
  • Technical cleanliness and contamination analysis
  • Inclusion and defect detection in metals and alloys
  • Coating and surface defect evaluation
  • Manufacturing process monitoring
  • Root-cause failure analysis
  • Steel and alloy inclusion classification
  • Corrosion and surface contamination analysis
  • Rapid failure investigation in production environments
Phenom ParticleX Desktop SEMs for particle analysis
Thermo Scientific Phenom ParticleX Desktop SEMs are optimized for particle analysis to automatically detect, classify, and statistically evaluate particles and contaminants. These tools are used for technical cleanliness compliance, wear debris analysis, inclusion monitoring, and incoming material inspection. Automated workflows support high-throughput analysis and standards-based reporting, making them ideally suited to routine quality control and production monitoring.
Desktop SEM techniques for automotive materials analysis
  • Particle and inclusion analysis
  • Automated and statistical analysis
  • Elemental screening
Automotive use cases for desktop SEM
  • Technical cleanliness compliance (ISO 16232, VDA 19.1)
  • Wear debris and contamination monitoring
  • Incoming material inspection
  • Routine quality assurance in manufacturing
X-ray photoelectron spectroscopy systems
XPS systems provide surface-sensitive chemical analysis of automotive materials, delivering information on elemental composition and chemical state within the top nanometers of a surface. They are used to investigate corrosion, passivation layers, coatings, adhesion failures, paint defects, and surface contamination. XPS is also essential for depth profiling and interface analysis in multilayer automotive systems.
XPS techniques for automotive materials analysis
  • Surface analysis
  • Chemical state analysis
  • Depth profiling and layer analysis
Automotive use cases of XPS
  • Corrosion and oxidation studies
  • Coating and passivation layer evaluation
  • Paint and adhesion defect analysis
  • Surface contamination identification
  • Multilayer material characterization
Transmission electron microscopes
TEMs offer nanoscale and atomic-level analysis of automotive materials. They are used to study precipitates, grain boundaries, phase transformations, and defect structures that influence mechanical strength, durability, and performance. TEMs are particularly valuable for advanced alloy development, analyzing battery materials, additive manufacturing, and failure analysis.
TEM techniques for automotive material analysis
  • Nanoparticle and microstructural analysis
  • Phase and structure analysis
  • Crystallographic analysis
Automotive use cases of TEM
  • Advanced alloy and lightweight material development
  • Battery material and interface analysis
  • Additive manufacturing microstructure evaluation
  • Precipitate and defect characterization
  • High-resolution failure analysis
Focused ion beam scanning electron microscopes
FIB-SEMs combine focused ion beam milling and scanning electron microscopy for site-specific sample preparation and 3D analysis. These systems deliver precise cross-sectioning, depth profiling, and volume reconstruction of complex automotive materials. FIB-SEM platforms are widely used for failure analysis, multilayer coatings analysis, additive manufacturing qualification, and battery interface studies.
FIB-SEM techniques for automotive materials analysis
  • Sample preparation
  • Depth profiling and layer analysis
  • 3D microstructure and volume analysis
  • Surface and interface analysis
Automotive use cases of FIB-SEM systems
  • Site-specific failure analysis
  • Multilayer coating and paint system analysis
  • Battery electrode and interface studies
  • Additive manufacturing qualification
  • Cross-sectioning of complex components
Cross-technique workflows
Automotive materials analysis often requires the combined use of multiple analytical techniques to achieve reliable results. Imaging, elemental analysis, surface analysis, and particle characterization are frequently applied in sequence, with each technique building on the results of the previous step. For example, particle and inclusion analysis can be used to identify areas of interest, surface-sensitive methods can then characterize chemistry and contamination, and sample preparation techniques support high-resolution or subsurface investigation. These integrated workflows support efficient root-cause analysis, confident decision-making, and consistent analysis across research, quality control, and production environments.