Search
Catalog Number(s)
| Components | Molecular Weight | Concentration (mg/L) | mM |
|---|---|---|---|
| Inorganic Salts | |||
| Ammonium Nitrate (NH4NO3) | 80.0 | 1650.0 | 20.625 |
| Calcium Chloride (CaCl2) (anhyd.) | 111.0 | 332.2 | 2.9927928 |
| Magnesium Sulfate (MgSO4) (anhyd.) | 120.0 | 180.7 | 1.5058333 |
| Potassium Nitrate (KNO3) | 101.0 | 1900.0 | 18.811882 |
| Potassium Phosphate monobasic (KH2PO4) | 136.0 | 170.0 | 1.25 |
1. Murashige, T. and Skoog, F. (1962) Physiol. Plant. 15, 485.