NF2 Antibody (711496) in KD
Antibody specificity was demonstrated by siRNA mediated knockdown of target protein. PC-3 cells were transfected with NF2 siRNA and decrease in signal intensity was observed in western blot application (Fig a) using ABfinity™ Anti - NF2 Recombinant Rabbit Oligoclonal Antibody (Product ## 711496). Densitometric analysis of this western blot is shown in histogram (Fig b).
Antibody specificity was demonstrated by siRNA mediated knockdown of target protein. PC-3 cells were transfected with NF2 siRNA and decrease in signal intensity was observed in western blot application (Fig a) using ABfinity™ Anti - NF2 Recombinant Rabbit Oligoclonal Antibody (Product ## 711496). Densitometric analysis of this western blot is shown in histogram (Fig b).
Related Products
Related Images