Search
Endogenous Tubulin (red) was labeled with Mouse anti-Tubulin primary antibody (322500) (1 ug/ml) in HeLa human cervical carcinoma cells and visualized using Goat anti-mouse IgG (H+L) Superclonal? Secondary Antibody, Alexa Fluor® 647 conjugate (A28181) (0.4 ug/ml, 1:2500). Nuclei (blue) were stained with SlowFade® Gold Antifade Mountant using DAPI (S36938) (1:50).