Neural Media and Reagents for Cell Therapy
Gibco reagents for neural cell culture have been trusted and published by researchers for more than twenty-five years. This trusted family of products is available under the CTS product line to provide workflow solutions that will:
  • Reduce your burden in qualifying reagents during transition from research to clinical applications
  • More easily meet regulatory and quality requirements
Learn more about what CTS products offer
Most popular CTS neural cell culture products
Products to help you move your research from the bench to the clinic
We offer a collection of Cell Therapy Systems (CTS) products for cell therapy research and further manufacturing of cell-, gene- and tissue-based products. For more information about our product offerings, ordering information and customer support, see our Gibco CTS brochure.
 Download brochureRequest more information
CTS Neurobasal Media
Designed for maintenance and expansion of human embryonic or post-natal neural stem cells and primary neurons. Allows for pure populations of neuronal cells without the need for an astrocyte feeder layer.
View product details
CTS B-27 Supplement, XenoFree portfolio
Xenofree variations of the original, trusted B-27 Supplement. All animal origin components from the original formulation have been replaced with recombinant or human origin components.
View product details
CTS N-2 Supplement
Serum-free, chemically defined supplement for the growth and expression of post-mitotic neurons and tumor cells of neuronal phenotype. 
View product details
Additional reagents for NSCs
CELLstart CTS substrate is a fully defined, xenofree substrate for neural stem cell growth and expansion.  Dilute with DPBS CTS media with calcium and magnesium.
View product details
CTS Hibernate Media
Serum-free, nutrient basal media for short-term maintenance of CNS tissues and cells at ambient COconditions. Use CTS Hibernate-A  Medium for postnatal neurons and CTS Hibernate-E  Medium for embryonic neurons.
View product details
CTS Synth-a-Freeze media
Chemically defined, protein-free liquid cryopreservation medium intended for freezing and storing a variety of mammalian cell types including neural stem cells.
View product details ›
Gibco CTS Growth Factors
High quality growth factors and cytokines for t-cell and dendritic cell applications.
View product details