• The Gibco CTS product line offers the broadest platform of "cell therapy–ready” solutions for your workflow.
  • Enter the clinic knowing that your tailored solutions will help meet your regulatory, quality, and clinical requirements.
  • The CTS product line enables you to reduce your burden in qualifying reagents during your transition from research applications to clinical applications.
We offer a collection of Cell Therapy Systems (CTS) products for cell therapy research and further manufacturing of cell-, gene- and tissue-based products. You can learn more about our product offerings with our CTS brochure or by requesting more information:
Media for cell therapy
We offer cell therapy-grade media to support both feeder-dependent and feeder-free pluripotent stem cell culture including Gibco KnockOut and Essential 8 products. 
See below for the extended list of reagents needed for stem cell maintenance and expansion. 
CELLstart CTS is a xeno-free stem cell substrate which is diluted with DPBS CTS w/ Ca & Mg.
GlutaMAX-1 CTS is added to the KnockOut SR XenoFree CTS complete medium, and TrypLE Select CTS is an AOF reagent used for gentle cell dissociation.
CTS growth factors
  • CTS offers high-quality growth factors and cytokines for T-cell and Dendritic cell applications
  • High biological activity - more results with less protein
  • High purity - no interference from other proteins or contaminants
  • Manufactured under ISO 13485 Quality Standard
  • DMF is available on request
CTS support
Technical and regulatory support and product documentation
View support information ›