Search
Thermo Scientific Avizo 3D Software for imaging data visualization and analysis has been enhanced with Digital Rock Analysis (DRA) extensions, offering a near-equivalent, scalable, and supported solution equivalent to Thermo Scientific PerGeos Software, which is now discontinued.
Leveraging the advanced, dedicated technology previously available in PerGeos Software, the new Avizo 3D Software Digital Rock Analysis extensions provide unique, proven visualization, processing, and analysis capabilities for improved evaluation of reservoir quality and faster understanding of static and dynamic rock properties that impact production.
Avizo Software 3D Pro's XCore Profile extension is a dedicated workspace that enables co-visualization of whole-core CT, core photography, and petrophysical logs in a single environment. The XCore Profile extension also creates virtual logs based on whole-core CT data for further calibration and communication of how changes in rock type impact how petrophysical properties are measured down-hole and allows creation of template report for processing routine analysis.
Avizo Software 3D Pro's XModeling & Simulation extension provides many processing capabilities for digital rock analysis. It provides a workflow to generate and extract a pore network model (PNM) based on 3D image data. It provides insight into pore and throat size distributions as a natural outcome of producing a pore network model. It also offers a powerful suite of advanced models for rock property analysis and simulations to obtain properties such as absolute permeability, formation factor, thermal properties, molecular diffusion.
DECT Xtra (Dual Energy Computed Tomography) adds value to whole-core CT analysis workflows by providing you with actual density information that can be compared to density logs and used for further characterization of rock formations. This enables you to compare whole-core CT images across multiple formations and wells.
For Research Use Only. Not for use in diagnostic procedures.