tramp metal detection
Tramp metal is metallic scrap that may accidentally find its way onto conveyors and into bulk materials. Some examples of tramp metal include bucket teeth, manganese steel mantles, bore crowns, bar scrap, chains, and tools. Tramp metal can damage expensive crushers, conveyors and other process equipment. As a result, tramp metal can stop operations and cause lost production time. Here are some frequently asked questions (FAQs) and answers about tramp metal detectors.
Contact a sales representative
A:  Tramp metal is metallic scrap that may accidentally find its way onto conveyors and into bulk materials. Some examples of tramp metal include bucket teeth, manganese steel mantles, bore crowns, bar scrap, chains, and tools.
A:  Tramp metal can damage expensive crushers, conveyors, and other process equipment. As a result, tramp metal can stop operations and cause lost production time.
A:  Tramp metal detectors protect expensive crushers, conveyors, and other process equipment from damage by tramp metal. The Thermo Scientific Ramsey Oretronic Tramp Metal Detector, for example, is designed for belt conveyors moving coal, iron pellets, minerals, aggregates, and other bulk materials, and can detect all types of metallic scrap that conventional metal detectors may miss, including bucket teeth, manganese steel mantles, bore crowns, bar scrap, chains, and tools.
A:  Yes. Many tramp metal detectors, including the Oretonric Tramp Metal Detector, can detect tramp metal when buried in wet conductive materials.
A:  Most tramp metal detectors are designed for belt conveyors moving coal, iron pellets, minerals, cement, aggregates, and other bulk materials. In the case of Oretronic Tramp Metal Detectors, the system coils are made of impact-resistant polyvinyl chloride and designed to withstand stresses of up to ten times the force of gravity. The transmitter coil and dual-wound receiver coils mount on a support made of non-conductive materials. The transmitter coil is swing-mounted to protect the coil and support from being struck from oversized materials. A dual receiver coil is utilized to counteract interference from external sources, such as electric motors, lightning, and radio transmitters.
The control unit enclosure for the Oretronic IV-4 is made with reinforced fiberglass that conforms to NEMA-4X, IP66 dust, and water tight 432 mm (17 in) x 360 mm (14.2 in) x 184 mm (7.2 in). The Oretronic IV-6 control has painted mild steel that conforms to NEMA-4, IP66 dust, and water tight 687 mm (27 in) x 524 mm (20.6 in) x 230 mm (9.1 in).
A:  Tramp metal detectors should feature variable frequency to eliminate interference from other electrical equipment, and password protection for security.
A:  Oretronic Tramp Metal Detectors can be used on conveyors with a belt speed of 1.5 m/min to 550 m/min (5 ft/min to 1800 ft/min).
For additional details about tramp metal detectors, read the Thermo Scientific Oretronic IV Tramp Metal Detector Product Specification Sheet.
Tramp Metal Detectors Help Keep Discarded Tires Out of Landfills  
Because tires contain metal, the shredding process itself can damage costly shredding equipment. During the shredding process, a tramp metal detector provides an economical and reliable method of protecting expensive shredders, crushers, conveyors, and other equipment from potentially costly damage from wheel bolts, which reduces downtime and ensures the cement production process stays operational. 
Support
Invest in peace of mind with a Thermo Scientific service agreement. Ensure minimum downtime with known costs and priority response times. Let us tailor a plan to meet your needs. For parts, technical support, or field assistance, please call 800-445-3503.