HPLC and UHPLC Systems
Excellence built into each innovative LC system
Separate even the most complex samples with confidence. With an extensive portfolio of innovative Thermo Scientific HPLC and UHPLC systems, you can choose the best system for your application. Learn which system is right for you, and customize it to meet your separation needs.
Quick links
Reliable, flexible, and robust chromatography with unmatched performance
Whether you need a high-end 1500 bar system or are looking for a simple-to-use nanoLC, learn how our innovative Thermo Scientific Vanquish UHPLC platform, our proven UltiMate 3000 systems, and our sensitive nano-, capillary-, and micro-flow rate systems can help you separate your science from the status quo.
Regardless of which system configuration you choose, you will receive a highly integrated setup with optimized fluidic connections. All of our HPLC and UHPLC systems have multiple detector options, and use our Thermo Scientific Viper Fingertight connections. Viper fittings are easy to install, completely tool free, virtually zero-dead volume connections that hold up to 1500 bar/22,000 psi.
Additionally, you can streamline your lab with our easy-to-use Thermo Scientific Chromeleon Chromatography Data System (CDS) software. Chromeleon CDS is the preferred software for single-point intelligent control, unique data processing tools, and intelligent run automation functionality. We also offer integration solutions for other CDS packages.
HPLC and UHPLC systems categories
Experience uncompromised UHPLC without compromising performance, robustness or ease of use. These systems are the most versatile and flexible systems that can handle any application. The Vanquish platform is equipped with unique Thermo Scientific SmartFlow and SmartInject technology, ensuring precision metering, gradient accuracy, and retention time and peak area reproducibility. Vanquish systems push chromatographic boundaries, and separate your toughest analytical challenges with confidence.
Discover why UltiMate 3000 LC systems are the proven workhorses of countless laboratories. These systems provide reliable chromatography and performance for a variety of applications at all scales. While being highly customizable with flexible system configurations and detector options, you can expect a fully integrated system with Chromeleon CDS software.
Are your sample volumes limited but require high sensitivity? Choose our high throughput solutions for applications such as LC-MS proteomics, metabolomics, or intact protein analysis. Achieve effortless nano-flow with our fully integrated Thermo Scientific EASY-nLC 1200, or leverage the flexibility of nano-, capillary-, and micro-flow rates with the customizable UltiMate 3000 RSLCnano system.  Both systems integrate seamlessly with Thermo Scientific mass spectrometers.
Not sure which system is right for you?
Connect with an expert to help you decide.
Contact us
Monitor your system remotely from anywhere in the world
  • Connect your lab to boost productivity with secure remote access to your data and instruments
  • Real-time monitoring of your instrument with automatic email notifications
  • Store data in secure accounts and share with colleagues and collaborators
Learn more
Featured HPLC and UHPLC systems video
HPLC and UHPLC basics
Learn about the history and basics of HPLC and UHPLC.
LC Columns and Consumables Catalog
Access the latest version of our Chromatography Columns and Consumables Catalog to ensure that you're optimizing your applications! Our enhanced online version features a robust applications search tool, access to method development guides, application notebooks, and much more.
Download catalog ›