Semi-Preparative HPLC Systems
High-performance purifications
The Thermo Scientific UltiMate 3000 Binary Semi-preparative system is a workhorse that will always be ready to assist with your purification of natural product extracts, isolation of impurities, or other large-scale LC processes.
UltiMate 3000 Binary Semipreparative System configuration
System UltiMate 3000 Semi-Preparative System
Pump Biocompatible Binary Pump
HPG-3200BX
Autosampler WPS-3000TSL Semi-preparative
Column compartment TCC-3000SD
Automated fraction collector AFC
Compatible components for all UltiMate 3000 HPLC and UHPLC systems
Multiple Detector Options HPLC Columns Chromeleon CDS Software
  • Charged aerosol detector
  • Diode array detector
  • Electrochemical detector
  • Fluorescence detector
  • Mass spectrometry detection
  • Multiple wavelength detector
  • Refractive index detector
  • Variable wavelength detector
  • An extensive portfolio of columns
  • In a broad variety of inner diameters, lengths, particle sizes, and chemistries
  • Streamlines your data processing
  • Industry-leading ease of use and intelligent functionality
  • Unique user guidance and hardware automation features designed for UltiMate 3000 systems
Discover the best HPLC systems for your applications and budget
Discover the best HPLC system for your applications and budget
Use these tools to help you decide
HPLC Systems Selection Guide
Use this tool to rank the top HPLC instruments for your applications based on your responses to simple pull-down menus.
HPLC System Total Cost of Ownership Calculator
Use this tool to estimate your cost for purchasing and running Thermo Scientific HPLC and UHPLC systems. You can compare costs between instruments.
LC Method Transfer Calculator
Use this online calculator to transfer methods from HPLC to UHPLC conditions. Calculate the new gradient table, adjust sample volumes and run times if applicable.