Vanquish Simple Switch 2D-LC Systems
Two independent flow paths for more confidence and productivity.
Vanquish Simple Switch 2D-LC Systems
Seamlessly switch between a trap heart-cut 2D-LC system and a dual LC system using the innovative Thermo Scientific Vanquish Simple Switch 2D-LC System. This two-dimensional LC (LC-LC) system is an innovative, all-in-one system that uses the novel Thermo Scientific Vanquish Dual Split Sampler to enable users to seamlessly switch between both system capabilities without re-plumbing. Simply decide which task needs to be performed and this system supplies the confidence and productivity you need.  
Highlights
Two systems in one
The unique system configuration allows you to run both trap-based 2D-LC for difficult samples and switch to a dual LC system that uses two independent 1D flow paths to run two analyses, either identical or not, on a single system without re-plumbing.
Greater confidence for complex sample analysis
Run sensitive LC-LC methods by enriching the fractionated analytes on a trap column to enable low-concentration detection and prevent solvent incompatibility between detection methods; or run two complementary assays simultaneously on two separate flow paths to double throughput.
Increased sensitivity
The system enables enrichment of low-abundant analytes and removal of diluting or disturbing solvents from the eluent by trapping and pre-concentrating the analytes of interest on a trap column to increase sensitivity when used as a 2D-LC instrument.
Simplified method development
The second-dimension pump provides the flow for solvent modulation, to address solvent incompatibility between UV and MS methods or mobile phase incompatibility between different column chemistries to support multiple combinations of 1D and 2D methods.
Flexibility
The modularity of the Vanquish LC systems makes it easy to build a system to best meet your application requirements with the flexibility to upgrade from a standard HPLC instrument to a more advanced configuration when you need to address more challenging analyses.  
Easy and reliable tool-free connections
The Vanquish Simple Switch 2D-LC system is fitted with Thermo Scientific Viper fingertight fittings, which support near-zero dead volume connections and enable sharper peaks and tool-free setup.
What is the Vanquish Simple Switch 2D-LC System?
Our unique Vanquish Simple Switch 2D-LC system allows you to simply switch between a trap-based 2D-LC or dual LC.
This flow scheme illustrates how the three six-port valves can be configured to run the system for 2D-LC analysis with two columns with a trap column in series, or as a dual LC system with two columns in parallel. 
All-in-one system: LC-LC system with trap and solvent modulation (left) OR Dual LC system with two independent ¹D flow path in one system (right).
Videos
System specifications
The table below provides an overview of the Vanquish Simple Switch 2D-LC system specifications for standard setups. Specifications may differ based on the specific system components selected. See detailed Component Specifications for more information.
Category Vanquish Simple Switch 2D-LC System
System pressure, maximum 1000 or 1500 bar, depends on system configuration
Flow range 0–5 or 0-8 mL/min, depends on pumps selected
Pump options (default gradient delay volume)
Binary (35, 200µL) 
Quaternary (679 µL) 
Delay volume depends on pump(s) selected
Biocompatible Yes
pH range 2–12, chloride concentration up to 1 mol/L
System design Modular
Touchscreen user interface display compatible (optional) Yes, depends on system configuration
Solvent monitoring system compatible (optional) Yes, depends on system configuration
Customizable gradient delay volume to simplify method transfer Yes, with metering device
SmartInject technology for maximum injection precision and improved column lifetime Yes
Injection volume range
Standard: 0.01-25 µL  
Optional: 0.01-100 µL
Sample cooling Yes, 4-40°C
Sample capacity (standard) 
Based on 12 mm OD vials (≤1.5 mL) for comparison
4 racks of 54 vials (≤ 1.5 mL) each, other rack formats available 
+ capacity of 12 × 22.5 mm OD vials (≤ 10 mL) in the carousel
Sample extension available (optional) Yes, with Vanquish Charger
Column compartment thermostatting
Yes, 5-120°C 
Still air and forced air
Column compartment pre-heater
Active and passive 
Column compartment post-column cooler Yes
Compatible Detectors
Vanquish Charged Aerosol Detector 
Vanquish Diode Array Detector  
Vanquish Fluorescence Detector 
Vanquish Multiple Wavelength Detector 
Vanquish Variable Wavelength Detector
Refractive Index Detector 
Mass Spectrometer
Component specifications
Standard Vanquish Simple Switch 2D-LC systems are made up of the following components. Download the associated specifications sheet for more detail on each component. 
  Vanquish Horizon Simple Switch 2D-LC System
Vanquish Flex Simple Switch 2D-LC System
Pump
Download Specification Sheet
Vanquish Binary Pump H 
or 
Vanquish Binary Pump H 
and 
Vanquish Quaternary Pump F
Vanquish Binary Pump F 
or 
Vanquish Binary Pump F 
and 
Vanquish Quaternary Pump F 
Note: Binary pump recommended as ²D pump
Autosampler
Download Specification Sheet
Vanquish Split Sampler HT Vanquish Split Sampler FT
Extended sample capacity (optional)
Download Specification Sheet
Vanquish Charger  
(optional)
Column compartment
Download Specification Sheet
Vanquish Column Compartment H 
Virtually zero dead volume system connections
Download Specification Sheet
Viper UHPLC Fittings
Detectors
Visit our HPLC and UHPLC Detectors page
Contact us to get a quote or discuss an HPLC/UHPLC solution
Our general purpose product lines, including LC-MS products, are not intended for in vitro diagnostic purposes in accordance with our product documentation, manuals, and labels. They are designated for General Laboratory Use Only.
Our general purpose product lines have not been tested or validated for such applications and their use for in vitro diagnostic purposes may result in health and safety risks. They are designated for Research Use Only Applications.
For Research Use Only - Not For Use In Diagnostic Procedures.