Search
VALID™ Software (P/N 4382356) provides integrated workflow tools and experimental plans to validate and implement Applied Biosystems 5-dye AmpFℓSTR® PCR Amplification Kits and is compatible with Applied Biosystems instruments and software including the 7500 Real-Time PCR System, 3130 Series Genetic Analyzers and GeneMapper® ID Software.
To utilize VALID™ Software in your lab, you'll need a combination of the following software and instrumentation:
Real-Time PCR instrumentation and software
Capillary Electrophoresis instrumentation and Data Collection software
To utilize VALID™ Software in your lab, you'll need a computer with the minimum requirements:
| Computer System Requirements | |
|---|---|
| MINIMUM REQUIREMENT | Computer Intel® Pentium® III processor 540 MHz 512 MB RAM 2 GB hard disk (free space) Monitor 17-inch monitor 1280 pixels X 1024 pixels |
| RECOMMENDED | Computer Intel® Pentium® III processor faster than 733MHz 1 GB RAM 10 GB EIDE hard drive 20/48X IDE CD-ROM Monitor 19 - inch or larger monitor 1280 pixels X 1024 pixels |
Click here for a full list of product compatibility.
Click here for more information about VALID™ Software.