LC-MS Systems
Do more with our precision LC-MS systems
Use Thermo Scientific liquid chromatography mass spectrometry (LC-MS) systems to identify, characterize and quantify unknown and known compounds within complex matrices, including drug metabolites, environmental toxins and food additives. Our systems enable you to streamline your workflow and obtain more information from every run, from high throughput sample screening to discovering trace levels of unknowns.
Featured mass spectrometry products
TSQ Altis Triple Quadrupole Mass Spectrometer
TSQ Quantis Triple Quadrupole Mass Spectrometer
Featured LC-MS system categories
Thermo Scientific Triple Quadrupole LC-MS/MS systems combine full scan capacity with the sensitivity of selected ion and reaction monitoring modes for targeted compound analysis.
Thermo Scientific Orbitrap mass spectrometers are designed for novel compound discovery in proteomics, biopharma, and metabolomics, using applications like isobaric tags and low level PTM analysis. These mass spectrometers generate high-resolution and accurate-mass data, thereby enhancing sensitivity in trace analyte detection, characterization and quantitation, and comprehensive sample analysis.
Ion Trap LC-MS brings extreme full-scan sensitivity and the power of data-dependent MSn analysis to bear on the detection and rapid identification of low-level components in complex matrices. Up to six orders of linear quantitation are also available.
These economical high-performance MS systems have the capacity to keep up with even the fastest UHPLC applications. They are ideal for routine analysis and function in full-scan and selected ion monitoring (SIM) modes. Thermo Scientific Chromeleon software helps users to master MS with integrated tools and guides that make operating and controlling your LC-MS system easy and intuitive.
Enhance your versatility with these add-ons
Upgrade your Orbitrap Fusion Lumos Tribrid MS to even higher resolution (1M), additional dissociation capabilities (UVPD and ETD), and a unique internal calibration (IC).
  • UVPD - Helps fragment lipids, peptides and intact proteins
  • 1M – Improves structural elucidation and quantitation of isobaric compounds
  • Easy-ETD - Fragments multiply charged precursors including PTMs
  • Easy-IC - Internal calibration improves mass accuracy
Learn more
Resources
Brochures
Support