Explosive ordnance disposal (EOD) equipment for quick explosives and chemical weapons detection
Make informed decisions while downrange
The explosive ordnance disposal (EOD) landscape is evolving. Homemade, improvised explosives, chemical threats, and locally manufactured ordnance items by terrorist organizations are growing concerns. EOD teams need to quickly identify potential explosive threats and detect chemical weapons to make informed decisions downrange on the target. We offer handheld Raman spectroscopy and FTIR chemical identification analyzers built for EOD teams, first responders and the military that give them actionable insights to disarm improvised explosive devices.
Contact a sales representative
Popular explosive ordnance disposal solutions
Explosive ordnance disposal categories
Rapidly and accurately identify various explosives and their precursors. With the increase of improvised explosive device (IED) usage worldwide, precise determination of explosives has never been more important. Military forces who need to analyze potential explosive threats and preserve evidence turn to us for portable identification systems to counter these threats.
Quickly analyze and identify the contents of unlabeled spilled material and illegally dumped drums. Traditional hazmat analysis at a remote laboratory adds time and expense to the remediation process. Our handheld analyzers harness the power of Raman or FTIR spectroscopy to quickly verify the contents of drums, bags, bottles and more directly in the hazard zone.
Video spotlight: explosive ordnance disposal solutions
FTIR and Raman spectroscopy instruments acquire a spectral fingerprint of an unknown substance, and then compare the collected fingerprint against a reference library. Both methods are lab-proven, precise optical technologies offering distinct advantages in specific applications. Used together, FTIR and Raman spectroscopy provide a broader range of unknown substance identification—and better protection for the responder and the community.
Read the application note