Antibody-Production-Art-250
We offer keyhole limpet hemocyanin (KLH) and other carrier proteins to conjugate peptides and haptens through sulfhydryl, amine or carboxyl groups to elicit the production of specific antibodies against almost any molecule. We also offer adjuvants for effective immunization.
Popular products
77150-Imject-cBSA-PBS-210
77120-Imject-OVA-PBS-210
77140-Freunds-Adjuvant-210
Featured products for antibody production
Conjugate KLH to proteins, peptides, or other haptens to ensure immunogenicity and high-yield antibody production from the conjugated antigen or use our custom conjugation service 
Purified forms of cationized bovine serum albumin (cBSA) that has been cationized (positively-charged) for the preparation of hapten-carrier protein conjugates having enhanced immunogenicity compared to BSA.
Purified and activated forms of Blue Carrier™ Protein, a highly immunogenic and soluble hemocyanin carrier protein for conjugation to peptides and other haptens to elicit a strong immune response and obtain high-yield antibody production.
Purified and activated forms of ovalbumin (OVA) from egg whites for use as a carrier protein to prepare immunogens for antibody production or secondary target for screening.
Purified and activated forms of bovine serum albumin (BSA), a convenient, highly soluble protein for use as a carrier in preparing immunogens for antibody production from haptens and other non-immunogenic antigens.
Prepared Freund's and alum-based adjuvants to mix and emulsify with prepared immunogens for preparation and subcutaneous or intravenous injection and immunization of animals for efficient antibody production.
bulk-pod-showcase-270x195
Partner with us to scale-up your process or product development pipeline with bulk quantities of high-quality immunoassay reagents and biochemicals. We offer a variety of compounds, devices, and resins for your large-scale applications.  
Complete request form